Skip to Content
Merck
All Photos(1)

Documents

30320

Sigma-Aldrich

Cytidine 5′-triphosphate disodium salt hydrate

≥90.0% (HPLC)

Synonym(s):

5-CTP-Na2, CTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14N3Na2O14P3 · xH2O
CAS Number:
Molecular Weight:
527.12 (anhydrous basis)
Beilstein:
3582735
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

≥90.0% (HPLC)

form

solid

impurities

≤12% water

solubility

H2O: 50 mg/mL, clear, colorless

shipped in

wet ice

storage temp.

−20°C

SMILES string

O.[Na+].[Na+].NC1=NC(=O)N(C=C1)[C@@H]2O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]2O

InChI

1S/C9H16N3O14P3.2Na.H2O/c10-5-1-2-12(9(15)11-5)8-7(14)6(13)4(24-8)3-23-28(19,20)26-29(21,22)25-27(16,17)18;;;/h1-2,4,6-8,13-14H,3H2,(H,19,20)(H,21,22)(H2,10,11,15)(H2,16,17,18);;;1H2/q;2*+1;/p-2/t4-,6-,7-,8-;;;/m1.../s1

InChI key

IJMSOWKYYIROGP-LLWADOMFSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Cytidine 5′-triphosphate disodium salt hydrate has been used:
  • in the oligonucleotide synthesis for silver nanoclusters interaction studies
  • as an anion source for fluorescence emission and titration curve-analysis of hypocrellin A-zinc(II) complex
  • in fluorescence emission experiments of copper(II)–2,6-bis(2-benzimidazolyl)pyridine complex

Cytidine 5γ-triphosphate (CTP) is used to synthesize RNA by various RNA polymerases. Cytidine 5γ-triphosphate (CTP) is a substrate for CMP-neuNAc synthetase(s) which produce CMP-neuNAc used in sialylation reactions. As a substrate of CTP:phosphocholine cytidylyltransferase(s) (CT), CTP supports the formation of phosphatidylcholine via the formation of CDP-choline.

Biochem/physiol Actions

Cytidine 5′-triphosphate (CTP) is essential for the synthesis of DNA and is a mediator of many physiological processes in living systems. It is crucial for the dolichol phosphorylation, cholesterol transport and is involved in the allosteric regulation of aspartate transcarbamylase. It is also used in the treatment of disease related brain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Melissa A MacKinnon et al.
The Journal of biological chemistry, 284(11), 7376-7384 (2009-01-15)
Phosphatidylcholine is the major phospholipid in eukaryotic cells. There are two main pathways for the synthesis of phosphatidylcholine: the CDP-choline pathway present in all eukaryotes and the phosphatidylethanolamine methylation pathway present in mammalian hepatocytes and some single celled eukaryotes, including
J Vionnet et al.
Glycobiology, 9(5), 481-487 (1999-04-20)
CMP-beta-N-acetylneuraminic acid (CMP-neuNAc) is the substrate for the sialylation of glycoconjugates by sialyltransferases in microbes and higher eukaryotes. CMP-neuNAc synthetase catalyzes the formation of this substrate, CMP-neuNAc, from CTP and neuNAc. In this report we describe the purification of CMP-neuNAc
Ratiometric Detection of Adenosine-5?-triphosphate (ATP) and Cytidine-5?-triphosphate (CTP) with a Fluorescent Spider-Like Receptor in Water
Gupta AK, et al.
European Journal of Organic Chemistry, 2015(1), 122-129 (2015)
Selective fluorometric detection of pyrophosphate and stringent alarmone with copper (II)-2, 6-bis (2-benzimidazolyl) pyridine complex
Zhao XJ and Huang CZ
Biosensors And Bioelectronics, 30(1), 282-286 (2011)
A molecular logic gate for the highly selective recognition of pyrophosphate with a hypocrellin A-Zn (II) complex
Zhao X and Huang CZ
Analyst, 135(11), 2853-2857 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service