Skip to Content
Merck
All Photos(1)

Key Documents

290R-1

Sigma-Aldrich

MUC1 (EP85) Rabbit Monoclonal Primary Antibody

Synonym(s):

Mucin 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP85, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (290R-14)
vial of 0.1 mL concentrate Research Use Only (290R-14-RUO)
vial of 0.5 mL concentrate (290R-15)
vial of 1.0 mL concentrate (290R-16)
vial of 1.0 mL concentrate Research Use Only (290R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (290R-17-RUO)
vial of 1.0 mL pre-dilute ready-to-use (290R-17)
vial of 7.0 mL pre-dilute ready-to-use (290R-18)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (290R-18-RUO)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500 (concentrated)

isotype

IgG

control

breast, colon (and Associated adenocarcinomas)

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

Gene Information

human ... MUC1(4582)

General description

Mucin 1 (MUC1) is a transmembrane glycoprotein that protects epithelial cells from injury caused by external stimuli. In addition to this role, MUC1 is involved in cell-cell adhesion, proliferation, motility, invasion and survival. In epithelial cells, MUC1 expression is regulated by binding of TNFα to TNFR1 and activation of the NFαB pathway. In human skin, MUC1 is not expressed in normal epidermis but rather in pre-malignant and malignant conditions. MUC1 has also been expressed in the epidermis of psoriatic plaques of biopsies from patients diagnosed with psoriasis vulgaris. MUC1, in addition to being localized at the apical surface of some suprabasal keratinocytes, is also localized over the entire cell surface of some of these cells and some basal keratinocytes. Conversely, no MUC1 immunoreactivity is typically detected in the epidermis of normal skin. Reactive gastropathy is the second most common diagnosis made on gastric biopsies. Increased epithelial proliferation and modifications of epithelial cytokeratin profile, distinct from those of Helicobacter pylori gastritis, have been previously reported. Loss of MUC1, either patchy or complete, was noted in 67% of the cases. The overexpression of MUC1 was found in 78% of papillary thyroid carcinomas (PTC). Multivariate analysis has shown a strong association of MUC1 expression with the presence of the BRAF (V600E) mutation and lymph node metastasis in PTC. Lymph node metastasis was the most important risk factor of relapse. In a study of thymic epithelial tumors, MUC1 was closely associated with VEGF, p53 and micro-vessel density (MVD). A 94% positive rate of MUC1 expression in thymic carcinomas and 0% in type B3 thymomas, demonstrated a significant difference. The expression of MUC1 was closely related to the grade of malignancy in thymic epithelial tumors. Positive expression of MUC1 was significantly associated with poor outcome in this disease. MUC1 may be useful marker for differentiating thymic carcinoma from type B3 thymoma.5

Quality


IVD

IVD

IVD

RUO

Linkage

MUC1 Positive Control Slides, Product No. 290S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Florence Renaud et al.
Thyroid : official journal of the American Thyroid Association, 24(9), 1375-1384 (2014-07-12)
The incidence of papillary thyroid carcinoma (PTC) has increased over the past 30 years in Western countries. PTC is usually associated with a good prognosis, but there is a wide range of aggressiveness, and some patients develop distant metastasis and/or
Kyoichi Kaira et al.
Virchows Archiv : an international journal of pathology, 458(5), 615-620 (2011-01-22)
MUC1 is a transmembrane mucin that has been related to tumor progression and outcome in various malignancies. However, the clinical significance in thymic epithelial tumors remains unknown. We investigated the clinical significance of MUC1 expression in thymic epithelial tumors. Fifty-five
Xiang-xiang Zhan et al.
Endocrine pathology, 26(1), 21-26 (2015-01-24)
The incidence of thyroid cancer has appeared as an increasing trend globally, especially in Asian countries. In this study, the expression of mucin-1 (MUC1) and Thomsen-Friedenreich antigen, Galβ1-3GalNAcα1-R (CD176) was investigated by immunohistochemistry in papillary thyroid carcinomas (PTCs), which accounts
Enrique Arciniegas et al.
Histology and histopathology, 30(4), 453-463 (2014-10-22)
Mucin 1 (MUC1) is a transmembrane glycoprotein that protects epithelial cells from injury caused by external stimuli. In addition to this role, MUC1 is involved in cell-cell adhesion, proliferation, motility, invasion and survival. In epithelial cells, MUC1 expression is regulated
Mari Mino-Kenudson et al.
Archives of pathology & laboratory medicine, 131(1), 86-90 (2007-01-18)
Reactive gastropathy is the second most common diagnosis made on gastric biopsies. Increased epithelial proliferation and modifications of epithelial cytokeratin profile, distinct from those of Helicobacter pylori gastritis, have been previously reported. However, the evaluation of mucins, important components of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service