Skip to Content
Merck
All Photos(1)

Key Documents

2853

Sigma-Aldrich

Eosin B Solution

0.1% aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
NACRES:
NA.47

form

solution

IVD

for in vitro diagnostic use

concentration

0.1%

application(s)

hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]C(=O)c1ccccc1C2=C3C=C(C(=O)C(Br)=C3Oc4c(Br)c([O-])c(cc24)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C20H8Br2N2O9.2Na/c21-14-16(25)11(23(29)30)5-9-13(7-3-1-2-4-8(7)20(27)28)10-6-12(24(31)32)17(26)15(22)19(10)33-18(9)14;;/h1-6,25H,(H,27,28);;/q;2*+1/p-2

InChI key

GYYTYUGGVBYJHE-UHFFFAOYSA-L

Application

Used as a counterstain in Sigma Procedure 285 for Fetal Hemoglobin.

Components

Eosin B, 0.1%, aqueous solution. Sodium azide, 0.1% added as preservative

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nilay S Sethi et al.
Nature genetics, 52(2), 219-230 (2020-02-07)
Somatic alterations in cancer genes are being detected in normal and premalignant tissue, thus placing greater emphasis on gene-environment interactions that enable disease phenotypes. By combining early genetic alterations with disease-relevant exposures, we developed an integrative mouse model to study
Lucía Margarita Valenzuela-Salas et al.
Pharmaceutics, 13(1) (2021-01-13)
Nanomaterials quickly evolve to produce safe and effective biomedical alternatives, mainly silver nanoparticles (AgNPs). The AgNPs' antibacterial, antiviral, and antitumor properties convert them into a recurrent scaffold to produce new treatment options. This work reported the full characterization of a
Rocky Strollo et al.
Diabetologia, 58(12), 2851-2860 (2015-09-10)
Insulin is the most specific beta cell antigen and a potential primary autoantigen in type 1 diabetes. Insulin autoantibodies (IAAs) are the earliest marker of beta cell autoimmunity; however, only slightly more than 50% of children and even fewer adults

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service