Skip to Content
Merck
All Photos(1)

Documents

19924

Sigma-Aldrich

Pyroglutamate Aminopeptidase from Pyrococcus furiosus, recombinant from E. coli

7-13 mU (per vial)

Synonym(s):

Pyrase, Pyroglutamyl-Peptidase I, Pyrrolidone Carboxyl Peptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in E. coli

Quality Level

form

lyophilized

specific activity

7-13 mU (per vial)

mol wt

Mr ~28000

storage temp.

−20°C

Related Categories

Packaging

package of 0.01 Unit

Unit Definition

1 U corresponds to the amount of enzyme which hydrolyzes 1 μmol Pyroglutamate-p-nitroanilide per minute at pH 7.0 and 37 °C

Analysis Note

enzyme activity: the optimum temperature is 95-100 °C (the enzyme is stable up to 75 °C), the optimum pH is 6-9 (stable from pH 6-9). Inhibitors: PCMB, Hg+.

Other Notes

An enzyme that removes pyroglutamic acid (pGlu) from pGlu-peptide and proteins; employed in Edman degradation.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Mozdzanowski et al.
Analytical biochemistry, 260(2), 183-187 (1998-07-11)
For larger proteins, efficient deblocking prior to Edman sequencing is especially important to obtain quality, extended sequencing data which is limited by the stepwise accumulation of background from the random acid hydrolysis of the protein. Therefore, any portion that remains
Y Shimada et al.
Journal of biochemistry, 106(3), 383-388 (1989-09-01)
The cDNA clone of Geotrichum candidum (Geo.) lipase was isolated from the Geo. cDNA library by colony hybridization using 32P-labeled oligonucleotides corresponding to a partial amino acid sequence of this enzyme. The nucleotide sequence of the cDNA determined by the
Gorka Larrinaga et al.
Neuroscience letters, 383(1-2), 136-140 (2005-06-07)
We evaluated the subcellular distribution of four membrane-bound aminopeptidases in the human and rat brain cortex. The particulate enzymes under study--puromycin-sensitive aminopeptidase (PSA), aminopeptidase N (APN), pyroglutamyl-peptidase I (PG I) and aspartyl-aminopeptidase (Asp-AP)--were fluorometrically measured using beta-naphthylamide derivatives. Membrane-bound aminopeptidase
Noriko Higaki-Sato et al.
Journal of agricultural and food chemistry, 54(19), 6984-6988 (2006-09-14)
In order to determine pyroglutamic acid levels in plasma, we developed a method based on precolumn derivatization of the carboxyl group of pyroglutamic acid with 2-nitrophenylhydrazine. Eight-week-old male SD strain rats were administered 200 mg of an acidic peptide fraction
Adolfo Varona et al.
American journal of physiology. Renal physiology, 292(2), F780-F788 (2006-09-21)
Peptides play important roles in cell regulation and signaling in many tissues and are regulated by peptidases, most of which are highly expressed in the kidney. Several peptide convertases have a function in different tumor stages, and some have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service