Skip to Content
Merck
All Photos(1)

Key Documents

19899

Sigma-Aldrich

CHAPS 100 mM solution

Synonym(s):

CHAPS solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H58N2O7S
CAS Number:
Molecular Weight:
614.88
MDL number:
UNSPSC Code:
12161902
PubChem Substance ID:
NACRES:
NA.25

description

zwitterionic

Quality Level

form

liquid

mol wt

614.88 g/mol

concentration

100 mM in H2O

technique(s)

protein quantification: suitable

CMC

8 mM

transition temp

cloud point >100 °C

density

1.01 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

C[C@H](CCC(=O)NCCC[N+](C)(C)CCCS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)CC4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C

InChI

1S/C32H58N2O7S/c1-21(8-11-29(38)33-14-6-15-34(4,5)16-7-17-42(39,40)41)24-9-10-25-30-26(20-28(37)32(24,25)3)31(2)13-12-23(35)18-22(31)19-27(30)36/h21-28,30,35-37H,6-20H2,1-5H3,(H-,33,38,39,40,41)/t21-,22?,23-,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

UMCMPZBLKLEWAF-RFCNGIAKSA-N

General description

3-[(3-cholamidopropyl) dimethylammonio]-1-propanesulfonate (CHAPS) is a biocompatible surfactant. It is a derivative of cholic acid comprising a hydrophobic cholesterol-like skeleton possessing three hydroxyl groups and a zwitterionic aliphatic tail.

Application

CHAPS 100 mM solution has been used as a component of lysis buffer for the extraction of proteins from goat endometrial epithelial cells (EECs) and liver tissue.

Biochem/physiol Actions

3-[(3-cholamidopropyl) dimethylammonio]-1-propanesulfonate (CHAPS) exhibits the properties of both sulfobetaine-type surfactants and bile salts. It is involved in the protein extraction process for proteomic analysis and reconstitution of membrane proteins into lipid vesicles (proteoliposomes). CHAPS is broadly used in biochemistry for membrane protein and receptor isolation and purification. It is conveniently used in pharmaceuticals and medicines due to its biocompatibility, noncharged character, nondenaturing, and disaggregating properties.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Youichi Shinozaki et al.
PLoS biology, 7(5), e1000103-e1000103 (2009-05-08)
The ATP-gated P2X(4) receptor is a cation channel, which is important in various pathophysiological events. The architecture of the P2X(4) receptor in the activated state and how to change its structure in response to ATP binding are not fully understood.
Morvarid Shafiei et al.
Applied biochemistry and biotechnology, 182(4), 1444-1457 (2017-02-01)
Pseudomonas aeruginosa biofilm-related infections are the major cause of premature death in cystic fibrosis patients. Strategies to induce biofilm dispersal are of interest, because of their potential in preventing biofilm-related infections. Our previous work demonstrated that n-butanolic Cyclamen coum extract
Justyna Kolakowska et al.
Reproduction, fertility, and development, 29(11), 2255-2268 (2017-04-19)
Reproductive processes in domestic pigs have been studied extensively. Pigs are one of the main sources of meat for human consumption and are an established model for investigations into mammalian, including human, reproductive physiology. Studies of the uterus during early
James O'Connell et al.
Nature communications, 10(1), 5795-5795 (2019-12-21)
Tumour necrosis factor (TNF) is a cytokine belonging to a family of trimeric proteins; it has been shown to be a key mediator in autoimmune diseases such as rheumatoid arthritis and Crohn's disease. While TNF is the target of several
Horst Funken et al.
PloS one, 7(10), e46857-e46857 (2012-10-12)
The fucose binding lectin LecB affects biofilm formation and is involved in pathogenicity of Pseudomonas aeruginosa. LecB resides in the outer membrane and can be released specifically by treatment of an outer membrane fraction with fucose suggesting that it binds

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service