Skip to Content
Merck
All Photos(1)

Key Documents

19176

Sigma-Aldrich

DL-Buthionine-sulfoximine

≥99.0% (TLC)

Synonym(s):

DL-Buthionine (S,R)-sulfoximine, BSO, Buthionine sulfoximine, Butionine sulfoximine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O3S
CAS Number:
Molecular Weight:
222.31
Beilstein:
2367136
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Assay

≥99.0% (TLC)

form

powder or crystals

mp

215 °C

solubility

H2O: 50 mg/mL, clear to almost clear, colorless

storage temp.

2-8°C

SMILES string

CCCCS(=N)(=O)CCC(N)C(O)=O

InChI

1S/C8H18N2O3S/c1-2-3-5-14(10,13)6-4-7(9)8(11)12/h7,10H,2-6,9H2,1H3,(H,11,12)

InChI key

KJQFBVYMGADDTQ-UHFFFAOYSA-N

Related Categories

Application

DL-Buthionine-sulfoximine is suitable for use to:
  • examine whether the inhibition of glutathione by BSO enhances the apoptotic effect of estrogen on antihormone-resistant human breast cancer cells
  • investigate the effect of BSO on development of bovine embryos
  • inhibit GSH in several studies
  • investigate the effect of GSH synthesis on oocyte maturation
It may be used to inhibit GSH and evaluate the hepatotoxicity and nephrotoxicity of natural food colorants in the absence of GSH

Biochem/physiol Actions

DL-Buthionine-sulfoximine inhibits the biosynthesis of Glutathione (GSH) in liver and other peripheral organs. It does not have any effect on GSH in the CNS. It augments the antiproliferative action of reactive oxygen species (e.g., hydrogen peroxide), and agents that indirectly cause accumulation of reactive oxygen species (e.g., 2-methoxyestradiol, which increases intracellular superoxide anion).

Other Notes

Depletes glutathionine in isolated cells

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christine Syng-Ai et al.
Molecular cancer therapeutics, 3(9), 1101-1108 (2004-09-16)
Curcumin, a well-known dietary pigment derived from Curcuma longa, inhibited growth of several types of malignant cells both in vivo and in vitro. However, its mechanism of action still remains unclear. In this study, we have focused primarily on the
N Yamauchi et al.
Biology of reproduction, 61(3), 828-833 (1999-08-24)
The present study was conducted to examine effects of cysteamine in culture medium on progression of meiosis, glutathione (GSH) content, kinase activities (histone H1 kinase and mitogen-activated protein kinase), and male pronuclear formation after in vitro insemination of cumulus-denuded oocytes
M Takahashi et al.
Biology of reproduction, 49(2), 228-232 (1993-08-01)
The purpose of this investigation was to determine the effect of beta-mercaptoethanol (beta-ME) and cysteamine, low-molecular-weight thiol compounds, on the development and intracellular glutathione content of bovine embryos obtained by in vitro fertilization of in vitro-matured oocytes. Embryos developed to
Jong-Shyan Wang et al.
European journal of applied physiology, 95(4), 290-297 (2005-08-13)
Exercise is linked with intensity-dependent immune response. Intracellular redox status is important in programmed cell death. This study, by closely examining 18 sedentary men who exercised moderately and severely (ie. 60% and 80% VO2max, respectively) for 40 min, investigated how
Depletion of glutathione by inhibition of biosynthesis.
O W Griffith
Methods in enzymology, 77, 59-63 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service