Skip to Content
Merck
All Photos(1)

Key Documents

17944

Sigma-Aldrich

1α,25-Dihydroxyvitamin D2

≥97.0% (sum of vitamin and previtamin, HPLC)

Synonym(s):

1α,25-Dihydroxycalciferol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O3
CAS Number:
Molecular Weight:
428.65
Beilstein:
5635017
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.79
Pricing and availability is not currently available.

biological source

synthetic

Quality Level

Assay

≥97.0% (sum of vitamin and previtamin, HPLC)

form

crystals

color

white to light yellow

storage temp.

−70°C

SMILES string

C[C@H](\C=C\[C@H](C)C(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C3\C[C@@H](O)C[C@H](O)C3=C

InChI

1S/C28H44O3/c1-18(9-10-19(2)27(4,5)31)24-13-14-25-21(8-7-15-28(24,25)6)11-12-22-16-23(29)17-26(30)20(22)3/h9-12,18-19,23-26,29-31H,3,7-8,13-17H2,1-2,4-6H3/b10-9+,21-11+,22-12-/t18-,19+,23-,24-,25+,26+,28-/m1/s1

InChI key

ZGLHBRQAEXKACO-XJRQOBMKSA-N

Biochem/physiol Actions

Ergocalciferol (vitamin D2) is activated in vivo by hydroxylation to 25-hydroxyergocalciferol and 1α,25-dihydroxycalciferol. Activated Vitamin D2 molecules may be used in a wide range of studies to assess their effects on functions such as immune function and calcium homeostasis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joanna Wietrzyk et al.
Oncology research, 16(11), 517-525 (2008-03-01)
New vitamin D analogs are interesting candidates for anticancer treatment, including squamous cell carcinomas (SCCs), especially in combination with cytostatics. In order to evaluate the effect of combined application of cisplatin, imatinib, or docetaxel and new vitamin D analogs [PRI-1906:
Shigeaki Kato et al.
Current opinion in nephrology and hypertension, 16(4), 297-304 (2007-06-15)
Vitamin D has diverse biological actions, and consequently the mechanisms behind how it regulates gene transcription are diverse. Unlike its well described positive effects on gene transcription, little is known about how vitamin D induces transcriptional repression. Vitamin D-induced transcriptional
R A Henik et al.
Journal of the American Veterinary Medical Association, 215(11), 1644-1649 (2003-10-22)
Rickets was diagnosed in a 1-year-old cat with a history of weakness, osteopenia, and recurrent fractures. Processes causing rickets include vitamin D deficiency caused by inadequate, nutrition, lack of exposure to sunlight, defective metabolism of parent vitamin D to active
Michał Chodyński et al.
Steroids, 67(9), 789-798 (2002-07-19)
A series of analogs of 1,25-dihydroxyergocalciferol (1-4) was synthesized and screened for their antiproliferative activity in vitro. The structure of new analogs was designed based on biological activity of the previously obtained side-chain modified analogs of vitamin D(2) and D(3).
Joanna Wietrzyk et al.
Cancer chemotherapy and pharmacology, 62(5), 787-797 (2008-01-12)
Active and less toxic vitamin D analogs could be useful for clinical applications. In the present study, we evaluated the toxicity and antitumor effect of two new synthetic analogs of vitamin D, namely PRI-1906 [(24E)-24a-Homo-(1S)-1,25-dihydroxyergocalciferol] and its side-chain unsaturated homo

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service