Skip to Content
Merck
All Photos(1)

Documents

17247

Sigma-Aldrich

N-Dodecanoyl-DL-homoserine lactone

≥97.0% (HPLC), suitable for ligand binding assays

Synonym(s):

N-Lauroyl-DL-homoserine lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H29NO3
CAS Number:
Molecular Weight:
283.41
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

N-Dodecanoyl-DL-homoserine lactone, ≥97.0% (HPLC)

Assay

≥97.0% (HPLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to faint beige

mp

114-117 °C

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCC(=O)NC1CCOC1=O

InChI

1S/C16H29NO3/c1-2-3-4-5-6-7-8-9-10-11-15(18)17-14-12-13-20-16(14)19/h14H,2-13H2,1H3,(H,17,18)

InChI key

WILLZMOKUUPJSL-UHFFFAOYSA-N

Application

Application test: Induces violacein expression in a Chromobacterium violaceum mutant usually not able to produce homoserine lactones.

Biochem/physiol Actions

N-Dodecanoyl-DL-homoserine lactone (C12-HSL) is among a group of homoserine lactones that includes; N-Heptanoyl-DL-homoserine lactone (C7HSL), N-octanoyl-homoserine lactone (N-C8-HSL), N-Decanoyl-DL-homoserine lactone (N-C10-HSL), N-(3-oxodecanoyl) homoserine-L-lactone (3-oxo-C10 HSL), N-(3-oxododecanoyl)homoserine-L-lactone (3-oxo-C12-HSL), N-(3-Oxotetradecanoyl)-L-homoserine lactone (3-oxo-C14-HSL, N-(3-hydroxydecanoyl)-L-homoserine lactone, and N-(3-hydroxyoctanoyl)-L-homoserine lactone involved in the processes of bacterial quorum sensing. These N-acyl-homoserine lactones are used to study the processes and mechanisms of bacterial quorum sensing.
N-Lauroyl- DL-homoserine lactone is a member of N-acyl-homoserine lactone family. N-acylhomoserine lactones (AHL) regulate gene expression in gram-negative bacteria, such as Echerichia and Salmonella are involved in quorum sensing, cell to cell communication among bacteria. Some AHLs are potent chemoattractants for human immune cells such as neutrophils.
Quorum-sensing signal generation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sophie Uzureau et al.
Journal of proteome research, 9(6), 3200-3217 (2010-04-15)
Many pathogenic bacteria use a regulatory process termed quorum sensing (QS) to produce and detect small diffusible molecules to synchronize gene expression within a population. In Gram-negative bacteria, the detection of, and response to, these molecules depends on transcriptional regulators
Deisy G Carneiro et al.
Frontiers in microbiology, 11, 1459-1459 (2020-08-28)
Acyl-homoserine lactones (AHLs) are quorum sensing (QS) signaling molecules that mediate cell-to-cell communication in Gram-negative bacteria. Salmonella does not produce AHL, however, it can recognize AHLs produced by other species through SdiA protein modulating important cellular functions. In this work
Kazunori Gomi et al.
Infection and immunity, 74(12), 7029-7031 (2006-09-20)
Chromobacterium violaceum produces autoinducers, including homoserine lactones (HSLs), for genetic regulation. Among the seven HSLs derived from C. violaceum we evaluated, only C(12)-HSL stimulated the production of inflammatory cytokines in mammalian monocytic cell lines through the activation of the NF-kappaB
Daniel Wynn et al.
The Analyst, 143(19), 4774-4782 (2018-09-15)
There are an estimated 48 million cases of foodborne illness in the United States every year. In general, these illnesses are the result of unintentional contamination and improper food handling. Because bacterial contamination plays a major role in food spoilage
Frank Wilco Bartels et al.
Biophysical journal, 92(12), 4391-4400 (2007-03-27)
Intercellular communication by means of small signal molecules coordinates gene expression among bacteria. This population density-dependent regulation is known as quorum sensing. The symbiotic nitrogen-fixing bacterium Sinorhizobium meliloti Rm1021 possesses the Sin quorum sensing system based on N-acyl homoserine lactones

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service