Skip to Content
Merck
All Photos(1)

Key Documents

16040

Sigma-Aldrich

Pyrogallol

ACS reagent, ≥99%

Synonym(s):

1,2,3-Trihydroxybenzene, 2,3-Dihydroxyphenol, Pyrogallic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3(OH)3
CAS Number:
Molecular Weight:
126.11
Beilstein:
907431
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
technique(s):
HPLC: suitable
application(s):
pharmaceutical (small molecule)
bp:
309 °C (lit.)
vapor pressure:
10 mmHg ( 167.7 °C)

grade

ACS reagent

vapor density

4.4 (vs air)

vapor pressure

10 mmHg ( 167.7 °C)

Assay

≥99%

form

solid

technique(s)

HPLC: suitable

ign. residue

≤0.005% (as SO4)

bp

309 °C (lit.)

mp

131.0-135.0 °C
133-134 °C (lit.)

solubility

water: soluble

anion traces

chloride (Cl-): ≤10 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.0005%

application(s)

pharmaceutical (small molecule)

SMILES string

Oc1cccc(O)c1O

InChI

1S/C6H6O3/c7-4-2-1-3-5(8)6(4)9/h1-3,7-9H

InChI key

WQGWDDDVZFFDIG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Pyrogallol has been used for preparing endothelial cell extract for determining vitamin E content using reverse-phase HPLC. It has also been used for preparing liver extracts for retinol analysis by using HPLC.

Biochem/physiol Actions

Pyrogallol also referred to as 1,2,3-trihydroxybenzene inhibits the response to nitric oxide (NO) in the rat anococcygeus muscle.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Muta. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J T Jang et al.
The Journal of nutrition, 130(5), 1291-1296 (2000-05-10)
In view of evidence that nutritional status of iron and vitamin A may affect the other nutrient's metabolism, we used model-based compartmental analysis to examine effects of iron deficiency on whole-body vitamin A dynamics in rats. Weanling male Sprague-Dawley rats
Fufang Wu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 215, 354-362 (2019-03-11)
Theophylline (TP) and theobromine (TB) are the methyl derivatives of xanthine. The antioxidation of TP and TB as well as the effect of the antioxidation and activity of copper‑zinc superoxide dismutase (SOD) with TP and TB were investigated. The contents
A Martin et al.
Arteriosclerosis, thrombosis, and vascular biology, 17(3), 429-436 (1997-03-01)
Monocyte adhesion to human aortic endothelial cells (ECs) is one of the early events in the development of atherogenesis. ECs were used to investigate the role of vitamin E in human monocyte adhesion to ECs in vitro. ECs incubated with
J S Gillespie et al.
British journal of pharmacology, 99(1), 194-196 (1990-01-01)
1. The effects of pyrogallol and hydroquinone on the bovine retractor penis (BRP) and rat anococcygeus muscles to non-adrenergic, non-cholinergic (NANC) nerve stimulation have been examined. Both drugs at a concentration of 10(-4) M significantly reduced the response in the
M Buchweitz et al.
Food chemistry, 135(4), 3010-3019 (2012-09-18)
The formation of anthocyanin-metal chelates, exhibiting intense blue colours was monitored over a period up to 10 weeks. Evaluating normalised absorption spectra in the range of 580-700 nm and their proportion of the total area under the curve (AUC), provided

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service