Skip to Content
Merck
All Photos(1)

Documents

14621C

SAFC

EX-CELL® 620-HSF Serum-Free Medium for Hybridoma Cells

Low-protein, without L-glutamine, liquid, sterile-filtered, suitable for cell culture

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352207
NACRES:
NA.75

Quality Level

description

for research or for further manufacturing use

sterility

sterile-filtered

form

liquid

technique(s)

cell culture | hybridoma: suitable
cell culture | mammalian: suitable

components

NaHCO3: 1.6 g/L
HEPES: 7.5 mM
L-glutamine: no
glucose: 3.45 g/L

shipped in

ambient

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

EX-CELL 620-HSF is a low-protein (approximately 11 mg/L) serum-free medium which has been specifically developed for the long-term growth of hybridoma and related cells capable of expressing monoclonal antibodies and other protein products. The following hybridoma, lymphoma and myeloma cell lines have demonstrated sustained long-term, serum-free growth (> 15 passages) in EX-CELL 620-HSF: Sp2/0, Ag14, L243, VD-10, Hut 78, K562. Additionally, transformed Chinese Hamster Ovary (CHO) cells have demonstrated excellent growth and productivity when grown in this medium.

Legal Information

Available in USA, Canada, and Europe. Please check with your local supplier regarding availability and restrictions in other countries.
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service