Skip to Content
Merck
All Photos(1)

Key Documents

138M-1

Sigma-Aldrich

CD138/syndecan-1 (B-A38) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

B-A38, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (138M-14)
vial of 0.5 mL concentrate (138M-15)
bottle of 1.0 mL predilute (138M-17)
vial of 1.0 mL concentrate (138M-16)
bottle of 7.0 mL predilute (138M-18)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:200

isotype

IgG1

control

tonsil

shipped in

wet ice

storage temp.

2-8°C

visualization

membranous

Gene Information

human ... SDC1(6382)

General description

CD138, Syndecan 1, is a transmembrane heparan sulfate proteoglycan, present mainly in a variety of epithelial tissues, participating in cell proliferation, cell migration, intercellular communication, and cell-matrix interactions. In lymphoid tissues, CD138 is expressed in the late stages of B-cell differentiation with progression towards plasma cells. Anti-CD138 is a good marker to selectively identify plasma cells, benign or malignant, in lymphoid tissues CD138 is also expressed in a variety of epithelial cells.

Quality


IVD

IVD

IVD

RUO

Linkage

CD138/syndecan-1 Positive Control Slides, Product No. 138S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Oksanen et al.
Journal of clinical pathology, 58(4), 376-381 (2005-03-26)
Both Helicobacter pylori and gastro-oesophageal reflux disease (GORD) may cause inflammation in cardiac mucosa. Intestinal metaplasia (IM) is found more often in GORD associated inflammation than in inflammation caused by H pylori, especially in young individuals. To examine morphological differences
Said J, et al.
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 261-261 null
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 254-254 null
Said J, et al.
WHO Classification of Tumours of Haematopoietic and Lymphoid Tissue, 261-261 null
M Chilosi et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 12(12), 1101-1106 (2000-01-05)
Detection of abnormal numbers and/or distribution of bone marrow (BM) plasma cells (PCs) on trephine biopsies can be important in the differential diagnosis of multiple myeloma (MM) and other PC disorders. A variety of immunohistochemical markers can potentially improve the

Related Content

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Diagnostic immunohistochemistry overview highlights its importance in cancer diagnosis and detecting infectious diseases in modern clinical pathology.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service