Skip to Content
Merck
All Photos(1)

Key Documents

10580

Sigma-Aldrich

Anthracene

suitable for scintillation, ≥99.0% (GC)

Synonym(s):

Anthraxcene, Paranaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H10
CAS Number:
Molecular Weight:
178.23
Colour Index Number:
10790
Beilstein:
1905429
EC Number:
MDL number:
UNSPSC Code:
12171500
eCl@ss:
39011608
PubChem Substance ID:
NACRES:
NA.32

vapor density

6.15 (vs air)

vapor pressure

1 mmHg ( 145 °C)

Assay

≥99.0% (GC)

form

powder or crystals

autoignition temp.

1004 °F

sublimation residue

≤0.1%

bp

340 °C (lit.)

mp

210-215 °C (lit.)
214-216 °C

solubility

alcohols: soluble
benzene: soluble
chloroform: soluble
hydronaphthalenes: soluble
supercritical carbon dioxide: soluble

fluorescence

λex 251 nm; λem 401 nm in acetonitrile

suitability

suitable for scintillation

SMILES string

c1ccc2cc3ccccc3cc2c1

InChI

1S/C14H10/c1-2-6-12-10-14-8-4-3-7-13(14)9-11(12)5-1/h1-10H

InChI key

MWPLVEDNUUSJAV-UHFFFAOYSA-N

Gene Information

human ... CYP1A2(1544)

Looking for similar products? Visit Product Comparison Guide

Application

Anthracene has been shown to be soluble in a variety of binary and ternary mixtures of cyclohexanone, ethyl acetate, and methanol .

Quantity

λmax. 357 nm, log ε 3.92

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kayleigh Slater et al.
Cancers, 12(10) (2020-10-18)
Metastatic uveal melanoma (UM) is a rare, but often lethal, form of ocular cancer arising from melanocytes within the uveal tract. UM has a high propensity to spread hematogenously to the liver, with up to 50% of patients developing liver
Payam Payamyar et al.
Chimia, 67(4), 283-285 (2013-08-24)
We describe the challenges involved with extending the limited lateral size of two-dimensional polymers (2DPs). An amphiphilic monomer with three-fold symmetry is chosen to form an ideally tessellated monolayer at the air/water interface. Anthracene [4+4] photo-dimerization is chosen as the
Hyunjung Lee et al.
Inorganic chemistry, 51(20), 10904-10915 (2012-09-26)
The tendency of a Hg(II) ion to strongly quench fluorescence of potential fluorescent sensors is explored. Fluorescence measurements show the expected order of the chelation-enhanced fluorescence (CHEF) effect of Zn(II) > Cd(II) > Hg(II) ~ Cu(II), which is interpreted as
Jiang-Fei Xu et al.
Organic letters, 15(24), 6148-6151 (2013-11-19)
Dynamic covalent bonds supplied by reversible anthracene dimerization were combined with pillar[5]arene/imidazole host-guest interactions to construct double-dynamic polymers. Heating such polymers (in solution or as a gel) led to depolymerization by dissociation of either the host-guest complexes alone or the
Ling-Bao Xing et al.
Langmuir : the ACS journal of surfaces and colloids, 29(9), 2843-2848 (2013-02-12)
A new type of anthracene organogelator based on uracil was obtained using organic aromatic solvents, cyclohexane, DMSO, ethanol, and ethyl acetate. It was further characterized by field-emission scanning electron microscopy and transmission electron microscopy. Specifically, the resulting organogels were demonstrated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service