Skip to Content
Merck
All Photos(1)

Key Documents

10112302

Shmac 4 cell line

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, human prostate, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

human prostate

packaging

tube of 5 μg 10112302-DNA-5UG
pkg of vial of cells 10112302-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

See Lang SH, et.al (2006)

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

Cell Line Origin

Prostate cancer moderately well differentiated, early stage

Packaging

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

DNA Profile

STR-PCR Data:

Amelogenin: X,Y
CSF1PO: 12
D13S317: 11
D16S539: 11,14
D5S818: 12,13
D7S820: 7,10
THO1: 7,9
TPOX: 8
vWA: 17,19

Culture Medium

Stemline Keratinocyte Medium II (Sigma S0196) + Stemline Keratinocyte Growth Supplement (Sigma S9945) + 2 mM Glutamine + 2% Fetal Bovine Serum (FBS). Addition of 2% serum was found to increase cell viability.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 to 1:10 seeding at approximately 3 x 104 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C. It is important to inactivate the trypsin with an equal volume of trypsin inhibitor (the culture medium contains too low a serum content to inactivate the trypsin). To remove all traces of trypsin and trypsin inhibitor pellet the cells by centrifugation and resuspend the cells in fresh medium. Media change every second day.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shona H Lang et al.
In vitro cellular & developmental biology. Animal, 42(8-9), 273-280 (2006-12-14)
Three-dimensional epithelial culture models are widely used to emulate a more physiologically relevant microenvironment for the study of genes and signaling pathways. Prostate epithelial cells can grow into solid cell masses or acinus-like spheroids in Matrigel. To test if the
Immortalization of human prostate cells with the human papillomavirus type 16 E6 gene.
Norman J Maitland et al.
Methods in molecular medicine, 88, 275-285 (2003-11-25)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service