Skip to Content
Merck
All Photos(1)

Documents

06966

Sigma-Aldrich

Atto 655-Biotin

BioReagent, suitable for fluorescence, ≥90% (HPCE)

Synonym(s):

Biotin-Atto 655

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116
NACRES:
NA.32

product line

BioReagent

Quality Level

Assay

≥90% (HPCE)

form

powder

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm
655 nm

solubility

DMF: soluble
DMSO: soluble
acetonitrile: soluble

fluorescence

λex 655 nm; λem 680 nm in 0.1 M phosphate pH 7.0

λ

in ethanol (with 0.1% trifluoroacetic acid: 655 nm ± 3 nm)

suitability

suitable for fluorescence

storage temp.

−20°C

Application

Atto 655 conjugated to biotin is useful for binding to proteins (i.e., avidin and streptavidin). The fluorescent properties include excellent thermal and photo-stability, high quantum yield, strong absorption (663 nm) and fluorescence (684 nm), and low triplet formation. It is a zwitterionic dye, carrying a net electrical charge of zero. Electron donors such as guanine and tryptophan can easily quench the fluorescence of this fluorophore. Biotin conjugated with ATTO 655 was applied to assess efficiency of BSA/avidin structures bound to a surface.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Julie M G Rogers et al.
Langmuir : the ACS journal of surfaces and colloids, 27(7), 3815-3821 (2011-03-16)
The structure and function of the influenza A M2 proton channel have been the subject of intensive investigations in recent years because of their critical role in the life cycle of the influenza virus. Using a truncated version of the
Ana J García-Sáez et al.
The Journal of biological chemistry, 286(43), 37768-37777 (2011-09-03)
Pore-forming toxins have evolved to induce membrane injury by formation of pores in the target cell that alter ion homeostasis and lead to cell death. Many pore-forming toxins use cholesterol, sphingolipids, or other raft components as receptors. However, the role
Søren Preus et al.
Chembiochem : a European journal of chemical biology, 13(14), 1990-2001 (2012-09-01)
Förster resonance energy transfer (FRET) is a powerful tool for monitoring molecular distances and interactions at the nanoscale level. The strong dependence of transfer efficiency on probe separation makes FRET perfectly suited for "on/off" experiments. To use FRET to obtain
Achim Friedrich et al.
FEBS letters, 581(8), 1644-1648 (2007-04-03)
This article presents a new, highly sensitive method for the identification of single nucleotide polymorphisms (SNPs) in homogeneous solutions using fluorescently labeled hairpin-structured oligonucleotides (smart probes) and fluorescence single-molecule spectroscopy. While the hairpin probe is closed, fluorescence intensity is quenched
Eilon Sherman et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 12(3), 696-703 (2011-01-29)
Intramolecular dynamics in the denatured state of a protein are of importance for protein folding. Native-like contact formation within the ensemble of denatured conformations of a protein may guide its transformation towards the native conformation. The efficiency of folding is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service