Skip to Content
Merck
All Photos(1)

Key Documents

05313

Sigma-Aldrich

L-Galactono-1,4-lactone

≥95.0% (GC)

Synonym(s):

L-Galactonic acid γ-lactone, NSC 25282

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
Beilstein:
83005
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95.0% (GC)

form

solid

optical activity

[α]/D 78.0±3.0°, c = 1 in H2O

color

white

mp

134  °C ((273 °F))

suitability

conforms to structure for Proton NMR spectrum

storage temp.

−20°C

SMILES string

OC[C@H](O)[C@H]1OC(=O)[C@@H](O)[C@@H]1O

InChI

1S/C6H10O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2-5,7-10H,1H2/t2-,3-,4-,5+/m0/s1

InChI key

SXZYCXMUPBBULW-NEEWWZBLSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Metabolite in the biosynthetic pathway of vitamin C in higher plants.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Schertl et al.
The Journal of biological chemistry, 287(18), 14412-14419 (2012-03-02)
L-galactono-1,4-lactone dehydrogenase (GLDH) catalyzes the terminal step of the Smirnoff-Wheeler pathway for vitamin C (l-ascorbate) biosynthesis in plants. A GLDH in gel activity assay was developed to biochemically investigate GLDH localization in plant mitochondria. It previously has been shown that
Targeting mitochondrial metabolism and machinery as a means to enhance photosynthesis.
Adriano Nunes-Nesi et al.
Plant physiology, 155(1), 101-107 (2010-10-23)
Joanna Maddison et al.
Planta, 214(3), 383-391 (2002-02-22)
Leaf L-ascorbate content of an ozone (O3)-sensitive radish genotype (Raphanus sativus L. cv. Cherry Belle) was increased 2-fold by feeding hydroponically cultivated plants L-galactono- 1,4-lactone (GalL). Plants were grown in controlled-environment chambers ventilated with charcoal/Purafil-filtered air, and administered one of
Rosalia Liso et al.
Journal of experimental botany, 55(408), 2589-2597 (2004-11-03)
To understand the function of ascorbic acid (ASC) in root development, the distribution of ASC, ASC oxidase, and glutathione (GSH) were investigated in cells and tissues of the root apex of Cucubita maxima. ASC was regularly distributed in the cytosol
Md Moinuddin Ahmed et al.
Carbohydrate research, 341(10), 1505-1521 (2006-04-18)
A short and highly efficient route to various C-4 substituted sugar lactones has been developed. The key to the overall transformation is the sequential osmium-catalyzed dihydroxylation reaction of substituted 2,4-dienoates and an allylic substitution at the C-4 position. When the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service