Skip to Content
Merck
All Photos(3)

Key Documents

C1396

Sigma-Aldrich

Calcium nitrate tetrahydrate

Calcium nitrate tetrahydrate
2 of 2 reviewers received a sample product or took part in a promotion

≥99.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(NO3)2 · 4H2O
CAS Number:
Molecular Weight:
236.15
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.55
Assay:
≥99.0%
form:
powder or crystals
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0%

form

powder or crystals

pH

5.5-7.0

mp

44 °C (dec.) (lit.)

storage temp.

room temp

SMILES string

O.O.O.O.[Ca++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Ca.2NO3.4H2O/c;2*2-1(3)4;;;;/h;;;4*1H2/q+2;2*-1;;;;

InChI key

ICSSIKVYVJQJND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Calcium nitrate tetrahydrate can be used as:
  • A template to prepare calcium(II)complexes of salicylaldimines derived from putrescine.
  • A catalyst in the solvent-free Biginelli three-component condensation reaction.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yogambha Ramaswamy et al.
Bioactive materials, 6(4), 1107-1117 (2020-10-27)
Surface topography is one of the key factors in regulating interactions between materials and cells. While topographies presented to cells in vivo are non-symmetrical and in complex shapes, current fabrication techniques are limited to replicate these complex geometries. In this
Mehdi Razavi et al.
Materials science & engineering. C, Materials for biological applications, 48, 21-27 (2015-01-13)
Although magnesium (Mg) is a unique biodegradable metal which possesses mechanical property similar to that of the natural bone and can be an attractive material to be used as orthopedic implants, its quick corrosion rate restricts its actual clinical applications.
Sang Heon Kim et al.
Analytica chimica acta, 1131, 80-89 (2020-09-16)
Sharp-tipped anisotropic silver (Ag) nanostructures are attracting increasing attention because of their unusual optical properties. However, the sharp tips make such nanostructures thermodynamically unstable; thus, they have been considered unsuitable for use in colorimetric sensing because of their tendency to
María Eugenia Goya et al.
Cell reports, 30(2), 367-380 (2020-01-16)
Recent discoveries have implicated the gut microbiome in the progression and severity of Parkinson's disease; however, how gut bacteria affect such neurodegenerative disorders remains unclear. Here, we report that the Bacillus subtilis probiotic strain PXN21 inhibits α-synuclein aggregation and clears
Kalimuthu Pandi et al.
Carbohydrate polymers, 134, 732-739 (2015-10-03)
Adsorption is a significant reaction occurs between adsorbent/water interface for controlling the pollutants in the aqueous environment. In this regard, an eco-magnetic biosorbent was prepared by uniform deposition of magnetic Fe3O4 particles on the surface of nano-hydroxyapatite (n-HAp)/chitosan (CS) nanocomposite

Questions

Reviews

2 of 2 reviewers received a sample product or took part in a promotion

Active Filters

  1. Miami, FL
    • Reviews 2
    • Vote 1
    5 out of 5 stars.

    For use in making a stock solution.

    The product was very easy to dissolve resulting in a clear solution.

    Helpful?

  2. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service