Skip to Content
Merck
All Photos(4)

Documents

A7302

Sigma-Aldrich

Ammonium molybdate tetrahydrate

ACS reagent, 81.0-83.0% MoO3 basis

Synonym(s):

Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)6Mo7O24 · 4H2O
CAS Number:
Molecular Weight:
1235.86
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

suitable for SM 5210

Assay

81.0-83.0% MoO3 basis

form

crystalline

reaction suitability

reagent type: catalyst
core: molybdenum

impurities

≤0.005% Insoluble matter

density

2.498 g/mL at 25 °C (lit.)

anion traces

arsenate, phosphate, and silicate (as SiO2): ≤0.001%
chloride (Cl-): ≤0.002%
nitrate (NO3-): passes test (limit about 0.003%)
phosphate (PO43-): ≤5 ppm
sulfate (SO42-): ≤0.02%

cation traces

Mg: ≤0.005%
heavy metals: ≤0.001% (as lead)

storage temp.

room temp

SMILES string

N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O

InChI

1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6

InChI key

FIXLYHHVMHXSCP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

Ammonium molybdate tetrahydrate (AMT) is a molybdenum salt, often employed in cryo-negative staining that helps to identify particles while preserving the material. As a catalyst, it is used to desulfurize thiophene and its derivatives.

Application

Ammonium molybdate tetrahydrate can be used as:
  • A molybdenum source in the synthesis of sodium molybdate dihydrate (Na2MoO4·2H2O).
  • A molybdenum source in the synthesis of nickel molybdate nanorods (NiMoO4) by co-precipitation method in the presence of nickel benzoate as a nickel source.
  • A precursor in the synthesis of Mn-MoS2 (MMS). MMS is employed in the production of MMS/rGO (reduced graphene oxide) composite that exhibits the highest rhodamine B (RhB) degradation efficiency.
  • A catalyst in the synthesis of PVDF (polyvinylidene fluoride) catalytic microcapsules. PVDF is a heterogeneous catalyst for the selective oxidation of benzyl alcohol to benzaldehyde.

Quality

May contain darkened ammonium molybdate crystals that are soluble in water.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian Ma et al.
Talanta, 88, 484-489 (2012-01-24)
Determination of silicate at low concentrations (i.e., nanomolar levels) is an important analytical objective for both marine science and the semiconductor industry. Here we report the use of flow injection analysis (FIA) in combination with long pathlength liquid core waveguide
Kyung Ho Park et al.
Journal of hazardous materials, 138(2), 311-316 (2006-07-25)
The petroleum refining industry makes extensive use of hydroprocessing catalysts. These catalysts contain environmentally critical and economically valuable metals such as Mo, V, Ni and Co. In the present study, a simple hydrometallurgical processing of spent hydrodesulphurization (HDS) catalyst for
Bernard Lorber et al.
Micron (Oxford, England : 1993), 39(4), 431-446 (2007-05-01)
The formation of 2D arrays of three small icosahedral RNA viruses with known 3D structures (tomato bushy stunt virus, turnip yellow mosaic virus and bromegrass mosaic virus) has been investigated to determine the role of each component of a negative
Pradeep Verma et al.
Bioresource technology, 102(4), 3941-3945 (2010-12-24)
Pretreatments for enzymatic saccharification are crucial for the establishment of lignocellulosic biorefineries. In this study, we focused on ammonium ions and peroxometal complexes as potential delignifying agents. We first examined the pretreatment of beech wood with nine different ammonium salts
Shifeng Cao et al.
International journal of food microbiology, 141(3), 173-176 (2010-07-08)
The potential enhancement of Pichia membranifaciens by ammonium molybdate (NH(4)Mo) to control blue mould caused by Penicillium expansum on peach fruit was investigated. Combining P. membranifaciens at 1x10(8) cell/ml with 1 mM NH(4)Mo provided a more effective control of blue

Protocols

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

To measure glucose-6-phosphatase activity, the Taussky-Shorr method is used. This method is a spectrophotometric stop-rate determination assay that is measured at 660 nm.

See All

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service