Skip to Content
Merck
All Photos(8)

Documents

746487

Sigma-Aldrich

Cesium chloride

anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99.9%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsCl
CAS Number:
Molecular Weight:
168.36
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

grade

anhydrous

Quality Level

product line

ReagentPlus®
Redi-Dri

Assay

99.9%

form

powder or crystals

quality

free-flowing

pH

6-7.5

mp

645 °C (lit.)

SMILES string

[Cl-].[Cs+]

InChI

1S/ClH.Cs/h1H;/q;+1/p-1

InChI key

AIYUHDOJVYHVIT-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Cesium chloride is a cesium halide. Cesium halides can be prepared by reacting cesium carbonate with corresponding hydrohalic acids. Cesium chloride ultracentrifugation method has been reported for the extraction of RNA from cellular fractions.

Application

Cesium chloride is used for the preparation of electrically conducting glasses. It is also used to make solutions for the separation of RNA from DNA by density gradient centrifugation.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sourav Saha et al.
Cell reports, 33(13), 108569-108569 (2020-12-31)
The present study demonstrates that topoisomerase 3B (TOP3B) forms both RNA and DNA cleavage complexes (TOP3Bccs) in vivo and reveals a pathway for repairing TOP3Bccs. For inducing and detecting cellular TOP3Bccs, we engineer a "self-trapping" mutant of TOP3B (R338W-TOP3B). Transfection with
Gloria Patricia Barrera et al.
BMC genomics, 16, 1008-1008 (2015-11-27)
Baculoviruses are insect-associated viruses carrying large, circular double-stranded-DNA genomes with significant biotechnological applications such as biological pest control, recombinant protein production, gene delivery in mammals and as a model of DNA genome evolution. These pathogens infect insects from the orders
Jie Wu et al.
Oncology reports, 35(1), 243-252 (2015-11-05)
Multigene-based combination therapy may be an effective practice in cancer gene therapy. Substantial studies have demonstrated that tumor suppressor p53 acetylation is indispensable for p53 activation. Inhibitor of growth 4 (ING4), as a novel tumor suppressor, is capable of remarkably
Ian C Jenkins et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(13), 4803-4808 (2014-03-19)
The degree to which DNA-linked particle crystals, particularly those composed of micrometer-scale colloids, are able to dynamically evolve or whether they are kinetically arrested after formation remains poorly understood. Here, we study a recently observed displacive transformation in colloidal binary
D R Alves et al.
Applied and environmental microbiology, 80(21), 6694-6703 (2014-08-26)
Biofilms are major causes of impairment of wound healing and patient morbidity. One of the most common and aggressive wound pathogens is Staphylococcus aureus, displaying a large repertoire of virulence factors and commonly reduced susceptibility to antibiotics, such as the

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service