Skip to Content
Merck
All Photos(9)

Key Documents

746460

Sigma-Aldrich

Lithium chloride

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

vapor pressure

1.33 hPa ( 547 °C)

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.008 meq/g Titr. base
≤0.01% insolubles

loss

≤1.0% loss on drying

pH

~6 (20 °C, 50 g/L)

mp

605 °C (lit.)

anion traces

sulfate (SO42-): ≤0.01%

cation traces

Ba: ≤0.003%
Ca: ≤0.01%
Fe: ≤0.001%
K: ≤0.01%
Na: ≤0.20%
heavy metals: ≤0.002% (by ICP-OES)

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium chloride (LiCl) is a lithium salt. It participates in the Horner-Wadsworth-Emmons reactions of base-sensitive aldehydes and phosphonates. LiCl/N,N-Dimethylacetamide (DMAc) solvent system has been employed for the dissolution of cellulose and synthesis of various cellulose derivatives (cellulose esters, carbamates, sulfonates and ethers). It can be synthesized from lithium carbonate (Li2CO3).

Application

Lithium chloride (LiCl) may be used in the synthesis of lithium perchlorate by reacting with perchloric acid.

Features and Benefits

  • Increased efficiency – Eliminate time and effort of loosening hardened powders
  • Safety – Avoid risks of using tools of force to break up clumps
  • Economy – Faster preparation and solvation increase productivity and reduce costs
  • Assured quality – Excellent, expert-tested quality with no anti-caking agents
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Na An et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(40), 14325-14331 (2014-09-17)
Human telomeric DNA consists of tandem repeats of the sequence 5'-TTAGGG-3' that can fold into various G-quadruplexes, including the hybrid, basket, and propeller folds. In this report, we demonstrate use of the α-hemolysin ion channel to analyze these subtle topological
Ted M Hsu et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(2), 327-337 (2014-07-19)
Glucagon-like peptide-1 (GLP-1) is produced in the small intestines and in nucleus tractus solitarius (NTS) neurons. Activation of central GLP-1 receptors (GLP-1Rs) reduces feeding and body weight. The neural circuits mediating these effects are only partially understood. Here we investigate
Alessandro Di Maio et al.
EvoDevo, 6, 17-17 (2015-07-15)
Wnt signaling is one of the earliest and most highly conserved regulatory pathways for the establishment of the body axes during regeneration and early development. In regeneration, body axes determination occurs independently of tissue rearrangement and early developmental cues. Modulation
Hector Vargas-Perez et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 34(23), 7899-7909 (2014-06-06)
Drug administration to avoid unpleasant drug withdrawal symptoms has been hypothesized to be a crucial factor that leads to compulsive drug-taking behavior. However, the neural relationship between the aversive motivational state produced by drug withdrawal and the development of the
Gang Chen et al.
Brain : a journal of neurology, 137(Pt 8), 2193-2209 (2014-06-13)
Accumulating evidence suggests that spinal cord astrocytes play an important role in neuropathic pain sensitization by releasing astrocytic mediators (e.g. cytokines, chemokines and growth factors). However, it remains unclear how astrocytes control the release of astrocytic mediators and sustain late-phase

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service