Skip to Content
Merck
All Photos(5)

Key Documents

650447

Sigma-Aldrich

2-Propanol

99.9%, HPLC Plus, for residue analysis, suitable for gas chromatography (GC), HPLC, suitable for ASTM® 7979 and ASTM® 7968

Synonym(s):

isopropanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
Beilstein:
635639
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:
NACRES:
NA.05
grade:
HPLC Plus
for residue analysis
technique(s):
HPLC: suitable
gas chromatography (GC): suitable
application(s):
PFAS testing
food and beverages
bp:
82 °C (lit.)
vapor pressure:
33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Product Name

2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%

grade

HPLC Plus
for residue analysis

Quality Level

Agency

suitable for ASTM® 7968
suitable for ASTM® 7979

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

99.9%

form

liquid

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.02% water
≤0.5 ppb Fluorescence (quinine) (365 nm)

evapn. residue

≤0.0001%

halogenated residue

≤10 ng/L (as heptachlor epoxide)

refractive index

n20/D 1.377 (lit.)

pH

7.0 (20 °C)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

solubility

water: soluble

density

0.785 g/mL at 25 °C (lit.)

λ

H2O reference

UV absorption

λ: 245 nm Amax: ≤1.0
λ: 250 nm Amax: ≤0.30
λ: 275-400 nm Amax: ≤0.005

suitability

suitable for residue analysis

application(s)

PFAS testing
food and beverages

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Propanol (Isopropanol) is a secondary alcohol. It has been tested as a substitute of the fuel for use in various fuel cells. CuO powder dissolved in 2-propanol has been used for the laser ablation assisted synthesis of Cu colloids. Suspension of 2-propanol with Zn(NO3)2 has been employed for the fabrication of titanium dioxide-coated stainless steel (P25-TiO2/SS) photoanode coated with uniformly thick layer of P25-TiO2. This photoanode was used for the electrochemical photocatalytic (ECPC) degradation process. Preparation of methyl isobutyl ketone (MIBK) using 2-propanol as the precursor has been reported with yield as high as 25%. Solubility of benzoic acid has been investigated in 2-propanol by gravimetric method in the temperature range, 277-346K.

Application

2-Propanol (Isopropanol) may be used in the following processes:
  • Isolation and quantitative determination of carotenes and xanthophylls from Capsicum annum pericarp extracts by HPLC.
  • As solvent in the dissolution of Pt and TiO2 precursors, for the fabrication of Pt-TiO2/Ti electrodes.
  • As a solvent in the synthesis of di- and trisubstituted ortho biaryls by Suzuki-Miyaura cross-coupling reaction.
  • As an alternate solvent to benzene in the analysis of total carbonyl compounds in heated and frying oils.
  • As a reaction medium in the preparation of cyclic ureas by reacting CO2 with diamines in the presence of pure cerium oxide (CeO2).
Suitable for HPLC, spectrophotometry, environmental testing

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Preparation Note

Product filtered through a 0.2 μm filter

Other Notes

Pure-Pac® II containers require the Micromatic MacroValve coupler for dispensing solvents, Z560723.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Pure-Pac is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical oxidation of alcohols on Pt-TiO2 binary electrodes.
Hasa B, et al.
International Journal of Hydrogen Energy, 38(35), 15395-15404 (2013)
A modified method for the estimation of total carbonyl compounds in heated and frying oils using 2-propanol as a solvent.
Endo Y, et al.
Journal of the American Oil Chemists' Society, 78(10), 1021-1024 (2001)
A general method for the Suzuki-Miyaura cross-coupling of sterically hindered aryl chlorides: Synthesis of di-and tri-ortho-substituted biaryls in 2-propanol at room temperature.
Navarro O, et al.
Journal of the American Chemical Society, 125(52), 16194-16195 (2003)
Highly efficient synthesis of cyclic ureas from CO2 and diamines by a pure CeO2 catalyst using a 2-propanol solvent.
Tamura M, et al.
Green Chemistry, 15(6), 1567-1577 (2013)
Solubility of benzoic acid in acetone, 2-propanol, acetic acid and cyclohexane: Experimental measurement and thermodynamic modeling.
Long B, et al.
Fluid Phase Equilibria, 297(1), 113-120 (2010)

Articles

Learn more about our Kava CRM solution for use in quantitative and qualitative analysis of kavalactones and flavokavains in kava products to confirm their potency and authenticity.

Learn more about our Kava CRM solution for use in quantitative and qualitative analysis of kavalactones and flavokavains in kava products to confirm their potency and authenticity.

Learn more about our Kava CRM solution for use in quantitative and qualitative analysis of kavalactones and flavokavains in kava products to confirm their potency and authenticity.

Learn more about our Kava CRM solution for use in quantitative and qualitative analysis of kavalactones and flavokavains in kava products to confirm their potency and authenticity.

Protocols

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

A guide to create solvent systems used for the thin-layer chromatography assay of Novabiochem products.

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service