Skip to Content
Merck
All Photos(1)

Key Documents

56678

Sigma-Aldrich

Hyflo® Super Cel®

filter aid, flux calcined, treated with sodium carbonate

Synonym(s):

Diatomaceous earth, flux-calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.52

form

solid

quality

flux calcined
treated with sodium carbonate

composition

Al2O3, 4.0%
CaO, 0.5%
Fe2O3, 1.5%
Na2O + K2O, 3.3%
SiO2, 89.6%

loss

~0.2% loss on ignition, 900 °C

median particle size

18 μm (acc. to granulometric method (laser sizer))

pH

>8.5 (25 °C, 10% in aq. suspension)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hyflo® Super Cel® (Diatomaceous earth), a commercial kieselguhr, is a promising substitute of quartz powder.

Application

Hyflo® Super Cel® may be used for the separation of mucopolysachharides by electrophoresis. It may be employed as filter aid during the preparation of protein renin isolated from horse plasma.

Legal Information

Hyflo is a registered trademark of Celite Corp.
Super Cel is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrophoresis of mucopolysaccharides in a slab of Hyflo Super Cel.
Gardell S, et al.
Acta Chemica Scandinavica, 4, 907-913 (1950)
L T SKEGGS et al.
The Journal of experimental medicine, 106(3), 439-453 (1957-09-01)
A purified preparation of a polypeptide renin substrate prepared by tryptic degradation of the protein renin substrate has been analyzed by the fluorodinitrobenzene method and after degradation with renin, carboxypeptidase, and phenylisothiocyanate, has been found to possess the amino acid
Myroslav Sprynskyy et al.
Journal of hazardous materials, 181(1-3), 700-707 (2010-06-15)
In this work the natural and the surfactant modified diatomite has been tested for ability to remove uranium ions from aqueous solutions. Such controlling factors of the adsorption process as initial uranium concentration, pH, contact time and ionic strength have
Guodong Sheng et al.
Environmental science & technology, 45(18), 7718-7726 (2011-08-23)
Sequestration of Ni(II) on diatomite as a function of time, pH, and temperature was investigated by batch, XPS, and EXAFS techniques. The ionic strength-dependent sorption at pH < 7.0 was consistent with outer-sphere surface complexation, while the ionic strength-independent sorption
Wenhui Xiong et al.
Journal of colloid and interface science, 368(1), 528-532 (2011-12-14)
This paper presents a novel technique integrating bulk-sensitive and surface-sensitive XANES methods to distinguish between physisorption and chemisorption for phosphate adsorption onto ferrihydrite-modified diatomite (FHMD). XANES P K-edge, L-edge, and Fe M-edge spectra were obtained for reference samples (K(2)HPO(4) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service