Skip to Content
Merck
All Photos(2)

Key Documents

538132

Sigma-Aldrich

2-Methyl-1-propanol

99.5%

Synonym(s):

Isobutanol, Isobutyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2OH
CAS Number:
Molecular Weight:
74.12
Beilstein:
1730878
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
Assay:
99.5%
bp:
108 °C (lit.)
vapor pressure:
8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

vapor density

2.55 (vs air)

Quality Level

vapor pressure

8 mmHg ( 20 °C)
8.8 mmHg ( 0 °C)

Assay

99.5%

form

liquid

autoignition temp.

801 °F

expl. lim.

10.6 %

impurities

≤0.05% (water)

refractive index

n20/D 1.396 (lit.)

bp

108 °C (lit.)

mp

−108 °C (lit.)

solubility

water: miscible 70 g/L at 20 °C

density

0.803 g/mL at 25 °C (lit.)

SMILES string

CC(C)CO

InChI

1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3

InChI key

ZXEKIIBDNHEJCQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methyl-1-propanol, also known as isobutyl alcohol, is an aliphatic alcohol. The densities, viscosities and refractive indices of the binary mixtures of diglyme and 2-methyl-1-propanol have been evaluated. The partition functions and thermodynamic properties of 2-methyl-1-propanol in the vapor phase have been reported. The kinetics of dehydration of isobutyl alcohol to butene over ZSM (Zeolite Socony Mobil)-5 zeolite has been investigated using in situ FTIR (Fourier transform infrared spectroscopy) and GC. The thermochemical characteristics of radicals generated by the removal of hydrogen from 1-butanol have been studied.

Application

2-Methyl-1-propanol may be used in the synthesis of corresponding ester by reacting with acetic acid in the presence of a solid superacid.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Densities, viscosities, and refractive indices of the binary mixtures of bis(2-methoxyethyl)ether with 1-propanol, 1-butanol, 2-methyl-1-propanol, and 2-methyl-2-propanol.
Aminabhavi TM and Gopalkrishna B.
Journal of Chemical and Engineering Data, 39(4), 865-867 (1994)
Mechanistic studies of the catalytic dehydration of isobutyl alcohol on NaH-ZSM-5.
Williams C, et al.
J. Chem. Soc., Faraday Trans., 86(20), 3473-3485 (1990)
Synthesis of Esters from Acetic Acid with Methanol, Ethanol, Propanol, Butanol, and Isobutyl Alcohol Catalyzed by Solid Superacid1.
Hino M and Arata K.
Chemistry Letters (Jpn), 10(12), 1671-1672 (1981)
Han Li et al.
Science (New York, N.Y.), 335(6076), 1596-1596 (2012-03-31)
One of the major challenges in using electrical energy is the efficiency in its storage. Current methods, such as chemical batteries, hydraulic pumping, and water splitting, suffer from low energy density or incompatibility with current transportation infrastructure. Here, we report
D Norbäck et al.
Occupational and environmental medicine, 52(6), 388-395 (1995-06-01)
As a part of the worldwide European Community respiratory health survey, possible relations between symptoms of asthma, building characteristics, and indoor concentration of volatile organic compounds (VOCs) in dwellings were studied. The study comprised 88 subjects, aged 20-45 years, from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service