Skip to Content
Merck
All Photos(1)

Documents

366374

Sigma-Aldrich

Zinc chloride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnCl2
CAS Number:
Molecular Weight:
136.30
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55

form

liquid

concentration

0.5 M in THF
470-600 mmol/L (silver nitrate titration)

density

0.948 g/mL at 25 °C

SMILES string

Cl[Zn]Cl

InChI

1S/2ClH.Zn/h2*1H;/q;;+2/p-2

InChI key

JIAARYAFYJHUJI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Zinc chloride solution is a colorless liquid that contains zinc chloride dissolved in THF. Its synthesis has been reported.

Application

Zinc chloride solution was used in the synthesis of the ferrocenylarenes.
It may be used for the in situ generation of organozincate reagents by reacting with Grignard compounds and also to catalyze chloromethylation of resins for solid phase peptide synthesis.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thin-layer electrochemistry of ferrocenylbenzene derivatives: Intramolecular electronic communication.
Wang MCP, et al.
Electrochimica Acta, 53(26), 7720-7725 (2008)
Zinc chloride-catalyzed chloromethylation of resins for solid phase peptide synthesis.
Feinberg RS and Merrifield RB.
Tetrahedron, 30(17), 3209-3212 (1974)
Versatile palladium (II)-catalyzed Negishi coupling reactions with functionalized conjugated alkenyl chlorides.
Peyrat JF, et al.
Tetrahedron Letters, 44(35), 6703-6707 (2003)
Russell E Carter et al.
Journal of neurochemistry, 125(5), 673-684 (2013-03-19)
Spreading depolarization (SD) is a feed-forward wave that propagates slowly throughout brain tissue and recovery from SD involves substantial metabolic demand. Presynaptic Zn(2+) release and intracellular accumulation occurs with SD, and elevated intracellular Zn(2+) ([Zn(2+) ]i ) can impair cellular
Molouk Beiromvand et al.
Preventive veterinary medicine, 109(1-2), 162-167 (2012-10-10)
Certain zoonotic parasites are enteropathogens in dogs that cause serious human disease such as cystic echinococcosis, human alveolar echinococcosis, visceral larva migrans, and ocular larva migrans. This study investigated the prevalence of intestinal parasites in dogs in the Chenaran County

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service