Skip to Content
Merck
All Photos(1)

Key Documents

34881

Sigma-Aldrich

Acetonitrile

≥99.8% (GC), suitable for HPLC-GC, gas chromatography (GC), HPLC

Synonym(s):

ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
Beilstein:
741857
EC Number:
MDL number:
UNSPSC Code:
41116105
eCl@ss:
39031501
PubChem Substance ID:
NACRES:
NA.03
Assay:
≥99.8% (GC)
technique(s):
HPLC-GC: suitable
HPLC: suitable
gas chromatography (GC): suitable
application(s):
food and beverages
bp:
81-82 °C (lit.)
vapor pressure:
72.8 mmHg ( 20 °C)

Product Name

Acetonitrile, suitable for HPLC-GC, ≥99.8% (GC)

vapor density

1.41 (vs air)

Quality Level

vapor pressure

72.8 mmHg ( 20 °C)

Assay

≥99.8% (GC)

form

liquid

autoignition temp.

973 °F

expl. lim.

16 %

technique(s)

HPLC-GC: suitable
HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.0005% non-volatile matter
≤0.001% free acid (as CH3COOH)
≤0.1% water (Karl Fischer)

transmittance

200 nm, ≥50%
220 nm, ≥90%
240 nm, ≥99%

refractive index

n20/D 1.344 (lit.)

bp

81-82 °C (lit.)

mp

−45 °C (lit.)

density

0.786 g/mL at 25 °C (lit.)

application(s)

food and beverages

format

neat

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Acetonitrile (MeCN) may be used as an extraction solvent during the quantification of pesticide residues in fruits and vegetables.
Used in the method development for pain panel and illicit drugs in clinical urine samples by LC-MS/MS.
It may be used as an extraction solvent during the quantification of pesticide residues in fruits and vegetables.

Other Notes

Important notice
  • The article number 34881-4X2.5L will be discontinued. Please order the single bottle 34881-2.5L which is physically identical with the same exact specifications.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Michelangelo Anastassiades et al.
Journal of AOAC International, 86(2), 412-431 (2003-05-02)
A simple, fast, and inexpensive method for the determination of pesticide residues in fruits and vegetables is introduced. The procedure involves initial single-phase extraction of 10 g sample with 10 mL acetonitrile, followed by liquid-liquid partitioning formed by addition of
Timo Glatter et al.
Molecular systems biology, 5, 237-237 (2009-01-22)
Protein complexes represent major functional units for the execution of biological processes. Systematic affinity purification coupled with mass spectrometry (AP-MS) yielded a wealth of information on the compendium of protein complexes expressed in Saccharomyces cerevisiae. However, global AP-MS analysis of
Elke H J Krekels et al.
Clinical pharmacokinetics, 53(6), 553-563 (2014-02-06)
From a previously validated paediatric population pharmacokinetic model, it was derived that non-linear morphine maintenance doses of 5 μg/kg(1.5)/h, with a 50 % dose reduction in neonates with a postnatal age (PNA) <10 days, yield similar morphine and metabolite concentrations
Hélène Hall et al.
Brain : a journal of neurology, 137(Pt 9), 2493-2508 (2014-07-27)
The neuropathological substrate of dementia in patients with Parkinson's disease is still under debate, particularly in patients with insufficient alternate neuropathology for other degenerative dementias. In patients with pure Lewy body Parkinson's disease, previous post-mortem studies have shown that dopaminergic
Divya Subramonian et al.
Journal of proteome research, 13(9), 3905-3918 (2014-07-30)
SUMOylation is an essential posttranslational modification and regulates many cellular processes. Dysregulation of SUMOylation plays a critical role in metastasis, yet how its perturbation affects this lethal process of cancer is not well understood. We found that SUMO-2/3 modification is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service