Skip to Content
Merck
All Photos(1)

Documents

33147

Sigma-Aldrich

1,4-Dioxane

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)

Synonym(s):

Diethylene oxide, Dioxane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8O2
CAS Number:
Molecular Weight:
88.11
Beilstein:
102551
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
puriss. p.a.

Agency

USP/NF
reag. ISO
reag. Ph. Eur.

vapor density

3 (vs air)

vapor pressure

27 mmHg ( 20 °C)
40 mmHg ( 25 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

356 °F

contains

~25 mg/L 2,6-di-tert.-butyl-4-methylphenol as stabilizer

expl. lim.

22 %

impurities

≤0.001% free acid (as CH3COOH)
≤0.001% non-volatile matter
≤0.005% acetaldehyde
≤0.005% peroxides (as H2O2)
≤0.01% carbonyl compounds (as HCHO)
≤0.05% water (Karl Fischer)

refractive index

n20/D 1.4210-1.4240
n20/D 1.422 (lit.)

pH

6.0-8 (20 °C, 500 g/L)

bp

100-102 °C (lit.)

mp

10-12 °C (lit.)

transition temp

solidification point ≤11 °C

density

1.034 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

C1COCCO1

InChI

1S/C4H8O2/c1-2-6-4-3-5-1/h1-4H2

InChI key

RYHBNJHYFVUHQT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,4-Dioxane is widely used as a solvent for various organic compounds in industry. It is a non-biodegradable contaminant found in industrial effluents. Oxidative degradation of 1,4-dioxane by using oxygen and ozone has been useful in converting it into biodegradable products, which can be easily removed by conventional methods.

Application

1,4-Dioxane has been employed as a solvent in the synthesis of N-(4-(prop-2-ynyloxy)phenyl)acetamide and 3-(4-(prop-2-ynyloxy)phenyl)-3,4-dihydro-2H-benzo[e][1,3]oxazine.

Other Notes

The article number 33147-6X1L will be discontinued. Please order the single bottle 33147-1L which is physically identical with the same exact specifications.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidation and biodegradability enhancement of 1,4-dioxane using hydrogen peroxide and ozone.
C D Adams et al.
Environmental science & technology, 28(11), 1812-1818 (1994-10-01)
Self-curable benzoxazine functional polybutadienes synthesized by click chemistry.
Kukut M, et al.
Designed Monomers and Polymers, 12(2), 167-176 (2009)
Christof Aellig et al.
ChemSusChem, 5(9), 1737-1742 (2012-07-05)
The dehydration of D-fructose to 5-hydroxymethylfurfural was studied under single-phase conditions in the low boiling solvent 1,4-dioxane at moderate temperatures in the presence of the solid acid-catalyst Amberlyst-15. The reaction was first examined and optimized under batch conditions, where it
Alkene trifluoromethylation coupled with C-C bond formation: construction of trifluoromethylated carbocycles and heterocycles.
Hiromichi Egami et al.
Angewandte Chemie (International ed. in English), 52(14), 4000-4003 (2013-02-27)
Xiaohua Liu et al.
Biomaterials, 31(2), 259-269 (2009-09-29)
It remains a challenge to synthesize functional materials that can develop advanced scaffolding architectures for tissue engineering. In this study, a series of biodegradable amphiphilic poly(hydroxyalkyl (meth)acrylate)-graft-poly(l-lactic acid) (PHAA-g-PLLA) copolymers have been synthesized and fabricated into nano-fibrous scaffolds. These copolymers

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service