Skip to Content
Merck
All Photos(1)

Key Documents

285234

Sigma-Aldrich

Bentonite

Synonym(s):

Montmorillonite

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
SB.52

form

powder

storage temp.

room temp

SMILES string

O.O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Al]O[Al]=O

InChI

1S/2Al.4O2Si.H2O.3O/c;;4*1-3-2;;;;/h;;;;;;1H2;;;

InChI key

KARVSHNNUWMXFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Bentonite is comprised primarily of the smectite group (montmorillonite) of clay minerals. It is widely used in various industrial applications such as clarification of edible and mineral oils, paints, cosmetics, and pharmaceuticals. It is also used as an adsorbent for the elimination of pollutants from wastewater.
The swelling property of bentonite is used to produce viscous water suspensions for bonding, plasticizing, and suspending applications. The presence of a large surface area of bentonite is due to the colloidal dispersion of the particles is the main reason for its application as an insecticide carrier, an emulsifier, and an emulsion stabilizer.

Application


  • Optimization of Cd (II) removal from aqueous solution by natural hydroxyapatite/bentonite composite using response surface methodology.: This study explores the use of a hydroxyapatite/bentonite composite for the efficient removal of cadmium ions from aqueous solutions, highlighting its potential application in environmental remediation (Desalegn et al., 2023).


  • Acid-activated Sodium Bentonite and Kaolin Clay: Comparative Study by Physicochemical Properties.: This research compares the physicochemical properties of acid-activated sodium bentonite and kaolin clay, focusing on their potential applications in various industrial processes and environmental treatments (Kumar & Lingfa, 2020).


  • Photochemical synthesis of silver nanoparticles on chitosans/montmorillonite nanocomposite films and antibacterial activity.: The research focuses on the synthesis of silver nanoparticles on chitosan/montmorillonite nanocomposite films and their enhanced antibacterial activity, suggesting potential applications in medical and environmental fields (Gabriel et al., 2017).



Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Applications of Bentonite in the Industrial Wastewater Treatment [J]
Xiaotong G and Zhiming Z
Chemical Production and Technology, 3 (2006)
Adsorption of methylene blue from aqueous solution by crosslinked chitosan/bentonite composite.
Bulut Y and Karaer H.
Journal of Dispersion Science and Technology, 36(1), 61-67 (2015)
Adsorption of phenol by bentonite
Banat FA, et al.
Environmental Pollution (Barking, Essex : 1987), 107(3), 391-398 (2000)
Laurent J Michot et al.
Langmuir : the ACS journal of surfaces and colloids, 29(10), 3500-3510 (2013-02-21)
The coagulation of sodium montmorillonite by inorganic salts (NaNO3, Ca(NO3)2 and La(NO3)3) was studied by combining classical turbidity measurements with wide-angle-X-ray scattering (WAXS), small-angle-X-ray scattering (SAXS), and transmission X-ray microscopy (TXM). Using size-selected samples, such a combination, associated with an
C D Wu et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(1), 152-158 (2012-11-07)
The single-solute and bisolute sorption behaviour of phenol and trichloroethylene, two organic compounds with different structures, onto cetyltrimethylammonium bromide (CTAB)-montmorillonite was studied. The monolayer Langmuir model (MLM) and empirical Freundlich model (EFM) were applied to the single-solute sorption of phenol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service