Skip to Content
Merck
All Photos(3)

Key Documents

251410

Sigma-Aldrich

Sodium thiocyanate

ACS reagent, ≥98.0%

Synonym(s):

Sodium isothiocyanate, Sodium rhodanate, Sodium rhodanide, Sodium sulfocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaSCN
CAS Number:
Molecular Weight:
81.07
Beilstein:
3594965
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0%
grade:
ACS reagent
form:
powder, crystals or chunks
solubility:
water: soluble 1,000 g/L at 20 °C
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Assay

≥98.0%

form

powder, crystals or chunks

impurities

≤0.005% insolubles
≤0.2% Na2CO3

pH

4.8 (20.1 °C, 1070 g/L)

mp

287 °C (dec.) (lit.)

solubility

water: soluble 1,000 g/L at 20 °C

anion traces

S2-: ≤0.001%
chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.01%

cation traces

Fe: ≤2 ppm
NH4+: ≤0.002%
heavy metals (as Pb): ≤5 ppm

SMILES string

[Na]SC#N

InChI

1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

VGTPCRGMBIAPIM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Undergoes nucleophilic substitution reactions with phase-transfer catalysts.[1]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sandmeyer trifluoromethylthiolation of arenediazonium salts with sodium thiocyanate and Ruppert-Prakash reagent.
Danoun G, et al.
Chemical Science, 5(4), 1312-1316 (2014)
Polym. Chem., 29, 243-243 (1991)
M-L Alvarez Gonzalez et al.
Human reproduction (Oxford, England), 24(9), 2217-2224 (2009-06-11)
In vivo mouse models have been developed to study the physiology of normal and pathologic endometrium. Although angiogenesis is known to play an important role in endometrial physiology and pathology, the origin of neovasculature in xenografts remains controversial. The aim
Catherine J Mitran et al.
Vaccines, 8(3) (2020-07-28)
In pregnant women, Plasmodium falciparum-infected red blood cells adhere to the placenta via the parasite protein VAR2CSA. Two vaccine candidates based on VAR2CSA are currently in clinical trials; however, these candidates failed to elicit strain-transcending antibody responses. We previously showed
Andrea Salis et al.
The journal of physical chemistry. B, 111(5), 1149-1156 (2007-02-03)
The effects of weak and strong electrolytes on the enzymatic activity of Candida rugosa lipase are explored. Weak electrolytes, used as buffers, set the pH, while strong electrolytes regulate the ionic strength. The interplay between pH and ionic strength has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service