Skip to Content
Merck
All Photos(1)

Documents

24127

Sigma-Aldrich

tert-Butanol

≥99% (GC)

Synonym(s):

2-Methyl-2-propanol, tert-Butyl alcohol, Trimethyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COH
CAS Number:
Molecular Weight:
74.12
Beilstein:
906698
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.5 (vs air)

vapor pressure

31 mmHg ( 20 °C)
44 mmHg ( 26 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

896 °F

expl. lim.

8 %

impurities

≤0.001% free acid (as C3H7COOH)
≤0.001% non-volatile matter
≤0.1% water (Karl Fischer)

refractive index

n20/D 1.387 (lit.)

pH

7 (20 °C)

bp

83 °C (lit.)

mp

23-26 °C (lit.)

density

0.775 g/mL at 25 °C (lit.)

SMILES string

CC(C)(C)O

InChI

1S/C4H10O/c1-4(2,3)5/h5H,1-3H3

InChI key

DKGAVHZHDRPRBM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butanol is a tertiary alcohol. Aqueous potassium peroxodisulfate undergoes photolysis in the presence of tert-butanol on irradiation with 254nm light at 20°C to afford hydrogen sulfate.

Application

tert-Butanol may be employed for the preparation of carbohydrate fatty acid esters. It may be employed as a reaction medium during the production of fatty acid methyl ester (FAME) from waste cooking palm oil (WCPO) by lipase-catalyzed reaction.

It can be used for the butylation of p-cresol in the presence of 12-tungstophosphoric acid supported on zirconia catalyst. 2-tert-Butyl-p-cresol (TBC), 2,6-di-tert-butyl-p-cresol (DTBC) and tert-butyl-p-tolyl ether (ether) are formed as butylation reaction products.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic studies of lipase on FAME production from waste cooking palm oil in a tert-butanol system.
Halim SFA and Kamaruddin AH.
Process. Biochem., 43(12), 1436-1439 (2008)
Sebastian Björklund et al.
Scientific reports, 7(1), 9960-9960 (2017-09-01)
Mesoporous silica has received much attention due to its well-defined structural order, high surface area, and tunable pore diameter. To successfully employ mesoporous silica for nanotechnology applications it is important to consider how it is influenced by solvent molecules due
Mojtaba Yegane Badi et al.
Journal of environmental health science & engineering, 17(2), 685-700 (2020-02-08)
The present study aimed to model and optimize the dimethyl phthalate (DMP) degradation from aqueous solution using UVC/ Na2S2O8/Fe2+ system based on the response surface methodology (RSM). A high removal efficiency (97%) and TOC reduction (64.2%) were obtained under optimum
The photolysis of potassium peroxodisulphate in aqueous solution in the presence of tert-butanol: a simple actinometer for 254nm radiation.
Mark G, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 55(2), 157-160 (1990)
Ilya Nifant'ev et al.
International journal of molecular sciences, 20(24) (2019-12-15)
There is a current clinical need for the development of bone void fillers and bioactive bone graft substitutes. The use of mesenchymal stem cells (MSCs) that are seeded into 3D scaffolds and induce bone generation in the event of MSCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service