Skip to Content
Merck
All Photos(3)

Key Documents

239321

Sigma-Aldrich

Sodium sulfite

ACS reagent, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SO3
CAS Number:
Molecular Weight:
126.04
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
form:
powder or crystals
solubility:
water: soluble 126 g/L at 20 °C
Pricing and availability is not currently available.

grade

ACS reagent

Agency

suitable for SM 5210

Assay

≥98.0%

form

powder or crystals

impurities

Free acid, passes test
≤0.005% insolubles
≤0.03 meq/g Titr. free base

pH

9.0-10.5 (25 °C, 126 g/L)

solubility

water: soluble 126 g/L at 20 °C

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.001%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].[Na+].[O-]S([O-])=O

InChI

1S/2Na.H2O3S/c;;1-4(2)3/h;;(H2,1,2,3)/q2*+1;/p-2

InChI key

GEHJYWRUCIMESM-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium sulfite is a water-soluble inorganic salt that is commonly used as a reducing agent. It is also used for sulfonation and sulfomethylation reactions. Additionally, it is used in the textile industry as a bleaching, de-sulfurizing, and dechlorinating agent. As an oxygen and free chlorine scavenger, Na2SO3 is employed in the water treatment process. Sodium sulfite participates in the purification and isolation of aldehydes and ketones by forming adducts with them. Other uses include the production of sodium thiosulfate, detergents, and dyes.

Application

Sodium sulfite can be used:
  • As a reducing agent in the hydroxylation of arylboronic acids and arylboronic esters into corresponding phenols.
  • In the transformation of α-amino alcohol methanesulfonate hydrochlorides to sodium α-amino alkanesulfonates.
  • As a SO2 precursor in the multigram synthesis of DABCO·(SO2)2 abbreviated as DABSO.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction kinetics of sodium sulfite oxidation by the rapid-mixing method.
Barron CH and O'Hern HA.
Chemical Engineering Science, 21(5), 397-404 (1966)
New plasticizers for wheat gluten films.
Irissin-Mangata J, et al.
European Polymer Journal, 37(8), 1533-1541 (2001)
Amber R Boles et al.
Biotechnology and bioengineering, 109(3), 637-646 (2011-10-22)
A novel sulfur-utilizing perchlorate reducing bacterial consortium successfully treated perchlorate (ClO₄⁻) in prior batch and bench-scale packed bed reactor (PBR) studies. This study examined the scale up of this process for treatment of water from a ClO ₄⁻ and RDX contaminated
Yisu Han et al.
ACS chemical biology, 14(4), 619-635 (2019-03-09)
APEX is an engineered peroxidase that catalyzes the oxidation of a wide range of substrates, facilitating its use in a variety of applications from subcellular staining for electron microscopy to proximity biotinylation for spatial proteomics and transcriptomics. To further advance
Qiang Yang et al.
Bioresource technology, 126, 336-340 (2012-11-06)
Agave americana is one of commonly grown agave species but currently less valuable because its large flower stalk cannot be used for producing alcoholic beverage. In the present study, the stalk was pretreated with dilute acid (DA), sulfite (SPORL), and

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service