Skip to Content
Merck
All Photos(2)

Key Documents

237078

Sigma-Aldrich

Aluminum chloride hexahydrate

ReagentPlus®, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3 · 6H2O
CAS Number:
Molecular Weight:
241.43
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.55
grade:
reagent
form:
crystalline solid
crystals or chunks

grade

reagent

vapor pressure

1 mmHg ( 100 °C)

product line

ReagentPlus®

Assay

99%

form

crystalline solid
crystals or chunks

reaction suitability

reagent type: catalyst
core: aluminum

pH

2.5-3.5 (20 °C)

SMILES string

Cl[Al](Cl)Cl.O.O.O.O.O.O

InChI

1S/Al.3ClH.6H2O/h;3*1H;6*1H2/q+3;;;;;;;;;/p-3

InChI key

JGDITNMASUZKPW-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum chloride hexahydrate is a nonflammable compound widely employed as a catalyst in organic synthesis. In addition to being extremely soluble in water, it is also soluble in alcohol, ether, glycerol, propylene glycol, and acetonitrile. It has many uses, including the refinement of crude oil, the production of deodorants and antiperspirant products, the dyeing of fabrics, textile finishing.

Application

Aluminum chloride hexahydrate is used as a catalyst:
  • In the synthesis of corresponding THP (tetrahydropyranylation) ether from alcohol or phenol, and 3,4-dihydro-2H-pyran (DHP).
  • In the direct esterification of levulinic acid and ethanol to produce ethyl levulinate.
  • In the synthesis of gem-dihydroperoxides from ketones and aldehydes.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Target Organs

Lungs

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark Baranov et al.
Inorganic chemistry, 58(13), 8877-8883 (2019-06-30)
While sophisticated computational methods can predict 31P NMR spectra of phosphorus atoms encapsulated within Keggin-derived heteropoly tungstate and molybdate cluster anions, calculated and experimental chemical shift values typically deviate considerably from one another. Motivated by the observation that experimentally determined
Ingrid Siemund et al.
Contact dermatitis, 77(5), 288-296 (2017-07-12)
Contact allergy to aluminium has been reported more frequently in recent years. It has been pointed out that positive patch test reactions to aluminium may not be reproducible on retesting. To investigate possible variations in patch test reactivity to aluminium
Aluminium (III) chloride hexahydrate: an efficient and versatile reagent in organic synthesis.
Sarma SD, et al.
ARKIVOC (Gainesville, FL, United States), 1, 243-263 (2013)
[Axillary hyperhydrosis. Local treatment with 25% aluminum chloride hexahydrate in absolute alcohol].
F Brandrup et al.
Ugeskrift for laeger, 140(35), 2106-2109 (1978-08-28)
A neutron and X-ray diffraction investigation of aluminum chloride hexahydrate.
Buchanan DR and Harris PM.
Acta Crystallographica Section B, Structural Science, 24(7), 954-960 (1968)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service