Skip to Content
Merck
All Photos(1)

Key Documents

22151

Sigma-Aldrich

Celite® 503

filter aid, treated with sodium carbonate, calcined

Synonym(s):

Diatomaceous earth, flux-calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.54

Quality Level

form

powder

quality

calcined
treated with sodium carbonate

composition

Al2O3, 4.0%
CaO, 0.5%
Fe2O3, 1.3%
Na2O + K2O, 3.3%
SiO2, 89.6%

loss

~0.2% loss on ignition, 900 °C

particle size

≤85.1 μm (80% (acc. to Cilas))

pH

>8.5 (25 °C, 10% in aq. suspension)

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Celite® 503 may be used as filter aid for the purification and isolation of crystalline desoxyribonuclease.

Caution

Not Intended for cGMP Parenteral Pharmaceutical Manufacturing

Legal Information

Celite is a registered trademark of Imerys Minerals California, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M KUNITZ
The Journal of general physiology, 33(4), 349-362 (1950-03-01)
A crystalline enzyme capable of digesting thymus nucleic acid (desoxyribonucleic acid) has been isolated from fresh beef pancreas. The enzyme called "desoxyribonuclease" is a protein of the albumin type. Its molecular weight is about 60,000 and its isoelectric point is
Gang Pan et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 13(8), 2205-2212 (2012-01-25)
The formation mechanism of a nanoscale gas state is studied on inorganic clay surfaces modified with hexamethyldisilazane, which show different contact angles in ethanol-water solutions. As the dissolved oxygen becomes oversaturated due to the decrease in ethanol-water ratio, oxygen nanoscale
Necla Caliskan et al.
Journal of hazardous materials, 193, 27-36 (2011-07-19)
The removal of Zn(II) ions from aqueous solution was studied using natural and MnO(2) modified diatomite samples at different temperatures. The linear Langmuir, Freundlich and Dubinin-Radushkevich (D-R) adsorption equations were applied to describe the equilibrium isotherms. From the D-R model
Guodong Sheng et al.
Environmental science and pollution research international, 20(6), 3708-3717 (2012-11-13)
This work determined the influence of humic acid (HA) and fulvic acid (FA) on the interaction mechanism and microstructure of Ni(II) onto diatomite by using batch experiments, X-ray photoelectron spectroscopy (XPS), and extended X-ray absorption fine structure (EXAFS) methods. Macroscopic
Baiyang Hu et al.
Journal of hazardous materials, 217-218, 85-91 (2012-04-03)
We developed a spongiform adsorbent that contains Prussian blue, which showed a high capacity for eliminating cesium. An in situ synthesizing approach was used to synthesize Prussian blue inside diatomite cavities. Highly dispersed carbon nanotubes (CNTs) were used to form

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service