Skip to Content
Merck
All Photos(2)

Key Documents

161551

Sigma-Aldrich

Activated charcoal

greener alternative

powder, -100 particle size (mesh), decolorizing

Synonym(s):

Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
11101522
PubChem Substance ID:
NACRES:
SB.52
Pricing and availability is not currently available.

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder

autoignition temp.

842 °F

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

resistivity

1375 μΩ-cm, 20°C (graphite)

particle size

-100 (mesh)

mp

3550 °C (lit.)

greener alternative category

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Activated charcoal (AC) powder, -100 particle size (mesh), decolorizing, is a carbonaceous material that is majorly used in a variety of applications due to its abundance, cost effectiveness and environmentally safe nature. It can be used in the fabrication of energy devices.[1]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

AC can be used as an electrode material in the formation of supercapacitors, double layered capacitors, lithium ion batteries and fuel cells.[1][2][3][4]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance studies of activated charcoal based electrical double layer capacitors with ionic liquid gel polymer electrolytes
Pandey GP, et al.
Energy & Fuels, 24(12), 6644-6652 (2010)
Fabrication of textile fabric counter electrodes using activated charcoal doped multi walled carbon nanotube hybrids for dye sensitized solar cells
Arbab AA, et al.
Journal of Material Chemistry A, 4(4), 1495-1505 (2016)
Supercapacitors from activated carbon derived from banana fibers
Subramanian V, et al.
The Journal of Physical Chemistry C, 111(20), 7527-7531 (2007)
Proton transfer reaction-mass spectrometry as a rapid inline tool for filter efficiency of activated charcoal in support of the development of Solid Oxide Fuel Cells fueled with biogas
Papurello D, et al.
Fuel Processing Technology, 130(12), 78-86 (2015)
Kishore Baathulaa et al.
Nature protocols, 6(12), 1990-1997 (2011-12-03)
Environment-sensitive fluorophores have different quantum yields in different solvents. 6-Dimethylaminonaphthalimide (6-DMN), for example, has a low quantum yield in aqueous solutions, but is highly fluorescent in nonpolar solvents or when bound to hydrophobic sites in proteins or membranes. 6-DMN has

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the surface area of this activated carbon?

    1 answer
    1. The surface area is not determined for this product.

      Helpful?

  4. Dear, could you please tell me if this activated charcoal is of vegetable or mineral origin? Is it a charcoal that has an acidic or basic pH?

    1 answer
    1. This product is not tested for pH. The source of this product is listed as 'Nature Organic'. Typically, this implies plant origin but may also include other sources such as lignite and coal. The specific source is considered proprietary and may vary from lot to lot. This information can be found on the lot-specific Certificate of Origin.

      Please navigate to the DOCUMENTATION section to access a Certificate:
      https://www.sigmaaldrich.com/product/sigald/161551#product-documentation

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service