Skip to Content
Merck
All Photos(3)

Documents

123226

Sigma-Aldrich

Anisole

ReagentPlus®, 99%

Synonym(s):

Methoxybenzene, Methyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H5
CAS Number:
Molecular Weight:
108.14
Beilstein:
506892
EC Number:
MDL number:
UNSPSC Code:
12352112
eCl@ss:
39023603
PubChem Substance ID:
NACRES:
NA.21

vapor density

3.7 (vs air)

Quality Level

vapor pressure

10 mmHg ( 42.2 °C)

product line

ReagentPlus®

Assay

99%

form

liquid

autoignition temp.

887 °F

expl. lim.

0.34-6.3 %

refractive index

n20/D 1.516 (lit.)

bp

154 °C (lit.)

mp

−37 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

SMILES string

COc1ccccc1

InChI

1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

RDOXTESZEPMUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Anisole can be obtained from the reaction between methyl formate and sodium phenolate or potassium phenolate at high temperature and pressure.

Application

Anisole has been used as a dopant to improve the sensitivity of photoionization at atmospheric pressure, which can be used as a source in the detection of nonpolar compounds using liquid chromatography coupled with mass spectrometry. It has also been used to improve the molecular fragmentation which further increases the background signal intensity in hyperthermal surface ionization source, when coupled with time-of-flight mass spectrometer.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adolfsson H, et al.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 37 (2014)
"A silica-based monolithic column in capillary HPLC and CEC coupled with ESI-MS or electrospray-atmospheric-pressure laser ionization-MS"
Droste S, et al.
Electrophoresis, 26(21), 4098-4103 (2005)
"Hyperthermal surface ionization in a time-of-flight mass spectrometer"
Weickhardt C and Draack L
Eur. J. Mass Spectrom., 6(4), 319- 323 (2000)
Adam F Lee et al.
Dalton transactions (Cambridge, England : 2003), 39(43), 10473-10482 (2010-10-05)
Size-controlled, catalytically active PVP-stabilised Pd nanoparticles have been studied by operando liquid phase XAS during the Suzuki cross-coupling of iodonanisole and phenylboronic acid in MeOH-toluene using KOMe base. XAS reveals nanoparticles are stable to metal leaching throughout the reaction, with
Shouhui Zhang et al.
The Journal of organic chemistry, 75(19), 6732-6735 (2010-09-09)
A CuI-catalyzed direct access to sulfides from disulfides via C-H bond cleavage of di- or trimethoxybenzene is described. The procedure utilizes O(2) as a clean and cheap oxidant. Direct selenation of the C-H bond also took place under this procedure.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service