Skip to Content
Merck
All Photos(1)

Key Documents

Y0001591

Flutamide for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Flutamide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11F3N2O3
CAS Number:
Molecular Weight:
276.21
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

flutamide

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)C(=O)Nc1ccc(c(c1)C(F)(F)F)[N+]([O-])=O

InChI

1S/C11H11F3N2O3/c1-6(2)10(17)15-7-3-4-9(16(18)19)8(5-7)11(12,13)14/h3-6H,1-2H3,(H,15,17)

InChI key

MKXKFYHWDHIYRV-UHFFFAOYSA-N

Gene Information

human ... AR(367)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Flutamide for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2 - STOT RE 2

Target Organs

Liver

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifeng Jing et al.
Cancer letters, 348(1-2), 135-145 (2014-03-26)
Androgen receptor (AR) has been indicated to be involved in bladder cancer progression. We showed androgen induced epithelial-mesenchymal transition (EMT) in AR-positive bladder cancer cells and promoted tumor metastasis in xenograft models. We subsequently identified that Slug was the mediator
F Labrie
Cancer, 72(12 Suppl), 3816-3827 (1993-12-15)
Although treatment of intact adult male rats with the pure antiandrogen flutamide or a luteinizing hormone-releasing hormone (LHRH) agonist alone leads to partial inhibition of ventral prostate weight, maximal inhibition is achieved by combination of the two drugs. Potentializing effects
R N Brogden et al.
Drugs, 38(2), 185-203 (1989-08-01)
Flutamide is a non-steroidal antiandrogenic drug devoid of hormonal agonist activity. Flutamide appears to be a specific antiandrogen only at androgen-dependent accessory genital organs. Its pharmacological activity is due substantially to the principal metabolite, 2-hydroxyflutamide. In comparative trials involving small
R Neri
Urology, 34(4 Suppl), 19-21 (1989-10-01)
Flutamide is rapidly metabolized by hydroxylation of the side chain to SCH 16423 (alpha, alpha, alpha-trifluoro-2-methyl-4'-nitro-m-lactotoluidide), the major metabolic product in all species studied, which is biologically active in vivo and in vitro studies. Flutamide exhibits its antiandrogenic activity by
Satoko Tange et al.
Environmental toxicology and pharmacology, 37(3), 996-1005 (2014-04-16)
Permethrin is a widely applied broad-spectrum pyrethroid insecticide that consists of a mixture of cis- and trans-isomers. We examined the changes of estrogenic and anti-androgenic activities resulting from metabolism of the isomers. Both cis- and trans-permethrin were hydrolyzed to 3-phenoxybenzyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service