Skip to Content
Merck
All Photos(1)

Key Documents

Y0001517

Haloperidol for system suitability

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Haloperidol, 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone, 4-[4-(p-Chlorophenyl)-4-hydroxypiperidino]-4′-fluorobutyrophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23ClFNO2
CAS Number:
Molecular Weight:
375.86
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

haloperidol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC1(CCN(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3

InChI

1S/C21H23ClFNO2/c22-18-7-5-17(6-8-18)21(26)11-14-24(15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2

InChI key

LNEPOXFFQSENCJ-UHFFFAOYSA-N

Gene Information

human ... HTR2A(3356)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Haloperidol for system suitability EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gerard W K Hugenholtz et al.
The Journal of clinical psychiatry, 67(6), 897-903 (2006-07-20)
To determine the doses of haloperidol as a comparator drug in randomized controlled trials (RCTs) with atypical antipsychotics in patients with schizophrenia and to compare these doses with the officially recommended doses for haloperidol in the United States and the
Jan Booij et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 55(4), 647-649 (2014-03-08)
A recent (123)I-FP-CIT ((123)-I-N-ω-fluoropropyl-2β-carbomethoxy-3β-(4-iodophenyl)nortropane) SPECT study on rats suggested that a single 1 mg/kg dose of the antipsychotic haloperidol induces enough dopamine release to compete with (123)I-FP-CIT for binding to the dopamine transporter. Taking into account the far-reaching consequences of
Mark C Fok et al.
International journal of geriatric psychiatry, 30(4), 333-344 (2015-02-03)
To summarize the effect of antipsychotics for preventing postoperative delirium. We conducted a literature search using Medline, EMBASE, Cochrane Central Register of Controlled Trials, Cochrane Database of Systematic Reviews and clinicaltrials.gov. We included randomized controlled trials of adults undergoing surgery
J M Goikolea et al.
Journal of affective disorders, 144(3), 191-198 (2012-10-24)
Treatment of acute mania with second-generation antipsychotics has been claimed to involve a lower risk of switch to depression than haloperidol. However, clinical guidelines clearly state that this is not a proven fact. Meta-analysis of double-blind randomized controlled trials in
Paul Perkins et al.
The Cochrane database of systematic reviews, (2)(2), CD006271-CD006271 (2009-04-17)
Nausea and vomiting are common symptoms of patients with terminal, incurable illnesses and can be distressing. The primary objective of the review was to evaluate the efficacy and adverse events associated with the use of haloperidol for the treatment of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service