Skip to Content
Merck
All Photos(1)

Key Documents

Y0001427

Carvedilol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(9H-Carbazol-4-yloxy)-3-[[2-(2-methoxyphenoxy)ethyl]amino]-2-propanol, BM-14190

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O4
CAS Number:
Molecular Weight:
406.47
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

carvedilol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC(COC1=CC=CC2=C1C3=C(C=CC=C3)N2)CNCCOC4=CC=CC=C4OC

InChI

1S/C24H26N2O4/c1-28-21-10-4-5-11-22(21)29-14-13-25-15-17(27)16-30-23-12-6-9-20-24(23)18-7-2-3-8-19(18)26-20/h2-12,17,25-27H,13-16H2,1H3

InChI key

OGHNVEJMJSYVRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Carvedilol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Cavedilol is a non-selective β-adrenergic blocker with α1 blocking activity. Carvedilol is used specifically for the treatment of heart failure and high blood pressure. It has been shown to improve left ventricular ejection fraction and may reduce mortality.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2 - STOT RE 2

Target Organs

Liver,spleen,Uterus (including cervix),Adrenal gland

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dhiraj Tripathi et al.
European journal of gastroenterology & hepatology, 22(8), 905-911 (2010-01-23)
Carvedilol is a potent noncardioselective beta-blocker, with weak vasodilating properties because of alpha 1 blockade. A reduction in both intrahepatic and portocollateral resistance contribute to enhanced effects on portal pressure. There are 10 published hemodynamic studies involving 168 patients investigating
Randall P Sharp et al.
The Annals of pharmacotherapy, 42(4), 564-571 (2008-03-28)
To examine the evidence regarding the impact of carvedilol on the serum lipid profile. Searches in MEDLINE and International Pharmaceutical Abstracts (1966-December 2007) were conducted. Search terms included carvedilol, cholesterol, lipids, hyperlipidemia, and beta-blockers. Published studies and case reports that
George L Bakris et al.
Reviews in cardiovascular medicine, 9(2), 96-105 (2008-07-29)
Few patients with hypertension meet recommended target blood pressure goals, and most hypertensive patients require at least 2 antihypertensive medications from different pharmacologic classes to adequately lower blood pressure. beta-Blockers are guideline-recommended for the treatment of hypertension with compelling indications.
Péter Ferdinandy et al.
Pharmacological reviews, 66(4), 1142-1174 (2014-09-28)
Pre-, post-, and remote conditioning of the myocardium are well described adaptive responses that markedly enhance the ability of the heart to withstand a prolonged ischemia/reperfusion insult and provide therapeutic paradigms for cardioprotection. Nevertheless, more than 25 years after the
Timothy Self et al.
The American journal of the medical sciences, 342(1), 56-61 (2011-02-05)
Cocaine-induced myocardial infarction (MI) is well documented. Current literature recommends avoiding beta-blockers in the acute care setting, but after discharge from the hospital, benefits of beta-blocker use may outweigh risks in patients with recent MI resulting from cocaine use. Cardioselective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service