Skip to Content
Merck
All Photos(1)

Key Documents

Y0001370

Olanzapine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Methyl-4-(4-methyl-1-piperazinyl)- 10H-thieno[2,3-b][1,5]benzodiazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4S
CAS Number:
Molecular Weight:
312.43
Beilstein:
7655141
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

olanzapine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CN1CCN(CC1)C2=Nc3ccccc3Nc4sc(C)cc24

InChI

1S/C17H20N4S/c1-12-11-13-16(21-9-7-20(2)8-10-21)18-14-5-3-4-6-15(14)19-17(13)22-12/h3-6,11,19H,7-10H2,1-2H3

InChI key

KVWDHTXUZHCGIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Olanzapine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jacqueline Flank et al.
Pediatric blood & cancer, 62(3), 496-501 (2014-10-21)
This retrospective review provides preliminary data regarding the safety and efficacy of olanzapine for chemotherapy-induced vomiting (CIV) control in children. Children <18 years old who received olanzapine for acute chemotherapy-induced nausea and vomiting (CINV) control from December 2010 to August
Susan L McElroy et al.
Current psychiatry reports, 17(5), 35-35 (2015-03-23)
Psychopharmacologic treatment is playing a greater role in the management of patients with eating disorders. In this paper, we review randomized, placebo-controlled trials (RCTs) conducted in anorexia nervosa (AN), bulimia nervosa (BN), binge eating disorder (BED), and other eating disorders
Hirotake Hida et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(3), 601-613 (2014-08-15)
Blonanserin differs from currently used serotonin 5-HT₂A/dopamine-D₂ receptor antagonists in that it exhibits higher affinity for dopamine-D₂/₃ receptors than for serotonin 5-HT₂A receptors. We investigated the involvement of dopamine-D₃ receptors in the effects of blonanserin on cognitive impairment in an
Karla Soares-Weiser et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 23(2), 118-125 (2012-05-29)
This comprehensive review and meta-analysis compared the effectiveness of olanzapine and other antipsychotics in schizophrenia treatment, defining effectiveness as time to all-cause medication discontinuation (primary) and as all-cause treatment discontinuation rates. This study examined randomized clinical trials (RCTs) and observational
Erin Schwenger et al.
Clinical pharmacokinetics, 50(7), 415-428 (2011-06-10)
Olanzapine, a second-generation antipsychotic, is a first-line agent in the treatment of schizophrenia. The objective of this review was to determine whether olanzapine warrants clinical pharmacokinetic monitoring in patients with schizophrenia, using a previously published decision-making algorithm. Although olanzapine is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service