Skip to Content
Merck
All Photos(1)

Key Documents

Y0001217

Piperine

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-Piperoylpiperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3
CAS Number:
Molecular Weight:
285.34
Beilstein:
90741
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

piperine

manufacturer/tradename

EDQM

mp

131-135 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C(\C=C\C=C\c1ccc2OCOc2c1)N3CCCCC3

InChI

1S/C17H19NO3/c19-17(18-10-4-1-5-11-18)7-3-2-6-14-8-9-15-16(12-14)21-13-20-15/h2-3,6-9,12H,1,4-5,10-11,13H2/b6-2+,7-3+

InChI key

MXXWOMGUGJBKIW-YPCIICBESA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Piperine EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yi Lin et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 3305-3310 (2013-11-26)
The aim of this study was to evaluate the cytotoxic and apoptotic effects of piperine on human lung cancer A549 cells and to explore its mechanisms. Piperine was found to exert the greatest cytotoxic effect against A549 cells in a
Angela Schöffmann et al.
Journal of medicinal chemistry, 57(13), 5602-5619 (2014-06-07)
Piperine activates TRPV1 (transient receptor potential vanilloid type 1 receptor) receptors and modulates γ-aminobutyric acid type A receptors (GABAAR). We have synthesized a library of 76 piperine analogues and analyzed their effects on GABAAR by means of a two-microelectrode voltage-clamp
Murlidhar Meghwal et al.
Phytotherapy research : PTR, 27(8), 1121-1130 (2013-04-30)
Black pepper (Piper nigrum L.) is a very widely used spice, known for its pungent constituent piperine. However, in addition to its culinary uses, pepper has important medicinal and preservative properties, and, more recently, piperine has been shown to have
Rohit A Gupta et al.
Journal of ethnopharmacology, 164, 239-246 (2015-02-17)
Piperine, a main component of Piper longum Linn. and Piper nigrum Linn., is a plant alkaloid with a long history of medicinal use. Piperine exhibits antidepressant, hepatoprotective, anti-metastatic, anti-thyroid, immunomodulatory, antitumor and anti-inflammatory activities, However its therapeutic potential in amelioration
Laia Rofes et al.
Journal of gastroenterology, 49(12), 1517-1523 (2013-12-12)
Oropharyngeal dysphagia (OD) is a major gastrointestinal motility disorder that causes severe nutritional and respiratory complications in elderly and neurological patients. In an earlier study, we found that stimulation of pharyngeal sensory neurons by capsaicinoids acting on transient receptor potential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service