Skip to Content
Merck
All Photos(1)

Key Documents

Y0001136

Lovastatin for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lovastatin, Mevinolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H36O5
CAS Number:
Molecular Weight:
404.54
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lovastatin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

−20°C

SMILES string

O1[C@@H](C[C@H](CC1=O)O)CC[C@@H]2[C@H]3[C@H](C[C@H](C=C3C=C[C@@H]2C)C)OC(=O)[C@H](CC)C

InChI

1S/C24H36O5/c1-5-15(3)24(27)29-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-19-12-18(25)13-22(26)28-19/h6-7,10,14-16,18-21,23,25H,5,8-9,11-13H2,1-4H3/t14-,15-,16-,18+,19+,20-,21-,23-/m0/s1

InChI key

PCZOHLXUXFIOCF-BXMDZJJMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lovastatin for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen-Huan Yu et al.
Carbohydrate polymers, 94(1), 56-62 (2013-04-03)
Two major fractions (RLP-1 and RLP-2) were obtained by purifying the crude polysaccharides extracted from a traditional Chinese herb Rosae Laevigatae Fructus. The average molecular weight of RLP-1 and RLP-2 was 21.5 kDa and 16.1 kDa, respectively. Monosaccharide analysis indicated
Emily K Osterweil et al.
Neuron, 77(2), 243-250 (2013-01-29)
Many neuropsychiatric symptoms of fragile X syndrome (FXS) are believed to be a consequence of altered regulation of protein synthesis at synapses. We discovered that lovastatin, a drug that is widely prescribed for the treatment of high cholesterol, can correct
Patricia A Reis et al.
PLoS pathogens, 8(12), e1003099-e1003099 (2013-01-10)
Cerebral malaria (CM) is the most severe manifestation of Plasmodium falciparum infection in children and non-immune adults. Previous work has documented a persistent cognitive impairment in children who survive an episode of CM that is mimicked in animal models of
Chia-Hao Chen et al.
International journal of pharmaceutics, 444(1-2), 18-24 (2013-01-29)
Lovastatin, categorized as a class II compound according to the Biopharmaceutics Classification System, is mainly responsible for the blood cholesterol lowering effect of red yeast rice (RYR). The aim of this study was to compare the dissolution rate, physical state
Kimberly A Hartwell et al.
Nature chemical biology, 9(12), 840-848 (2013-10-29)
Efforts to develop more effective therapies for acute leukemia may benefit from high-throughput screening systems that reflect the complex physiology of the disease, including leukemia stem cells (LSCs) and supportive interactions with the bone marrow microenvironment. The therapeutic targeting of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service