Skip to Content
Merck
All Photos(1)

Key Documents

Y0001034

Lynestrenol for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Lynestrenol, (17α)-19-Norpregn-4-en-20-yn-17-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H28O
CAS Number:
Molecular Weight:
284.44
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

lynestrenol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@@]1([C@@]2([C@H]([C@H]3[C@@H]([C@H]4CCCC=C4CC3)CC2)CC1)C)C#C

InChI

1S/C20H28O/c1-3-20(21)13-11-18-17-9-8-14-6-4-5-7-15(14)16(17)10-12-19(18,20)2/h1,6,15-18,21H,4-5,7-13H2,2H3/t15-,16+,17+,18-,19-,20-/m0/s1

InChI key

YNVGQYHLRCDXFQ-XGXHKTLJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Lynestrenol for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yasir Hasan Siddique et al.
Indian journal of experimental biology, 43(3), 291-293 (2005-04-09)
Genotoxicity study of synthetic progestin lynestrenol, was carried out on mouse bone marrow cells using sister chromatid exchanges (SCEs) and chromosomal aberrations (CAs) as parameters. Lynestrenol was studied at three different doses (6.87, 13.75 and 27.50 mg/kg body wt.). SCE
Ozlem Ozdegirmenci et al.
Gynecologic and obstetric investigation, 72(1), 10-14 (2011-01-27)
To evaluate the treatment of endometrial hyperplasia (EH) with different progestins. Eighty-two women with simple EH without atypia were included. Patients were offered oral progestagens and were randomized to one of three options for 3 months: medroxyprogesterone acetate (MPA, 10
Eva-Katrin Bentz et al.
Fertility and sterility, 94(7), 2688-2696 (2010-06-12)
To evaluate gene expression signatures of breast tissue in female-to-male (FtM) transsexuals under cross-sex hormone therapy (HT). Prospective cohort study. Academic research institution. Five hormone-naïve FtM transsexuals before and after HT. Breast tissue biopsy before and after 2 years of
m A Dzhangidze et al.
Georgian medical news, (145)(145), 21-25 (2007-05-26)
The effect of hormonal contraception on cervix of the uterus of young nullipara women with ectopia was investigated. Cohort study was carried out by using simple blind method. By means of statistical data manipulation correlation between the application of hormonal
Magali Svrcek et al.
European journal of endocrinology, 156(6), 617-621 (2007-05-31)
The relationship between sex hormones and hepatocellular adenoma development is well established. On the contrary, their contribution to liver adenomatosis (LA) development is still a debatable issue. Recently, inactivating mutations of hepatocyte nuclear factor-1alpha (HNF-1alpha) transcription factor gene or activating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service