Skip to Content
Merck
All Photos(1)

Key Documents

Y0000806

Arbutin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

4-Hydroxyphenyl-β-D-glucopyranoside, p-Arbutin, Hydroquinone β-D-glucopyranoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O7
CAS Number:
Molecular Weight:
272.25
Beilstein:
89673
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

arbutin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc(O)cc2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H16O7/c13-5-8-9(15)10(16)11(17)12(19-8)18-7-3-1-6(14)2-4-7/h1-4,8-17H,5H2/t8-,9-,10+,11-,12-/m1/s1

InChI key

BJRNKVDFDLYUGJ-RMPHRYRLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Arbutin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J J Leyden et al.
Journal of the European Academy of Dermatology and Venereology : JEADV, 25(10), 1140-1145 (2011-06-01)
Facial hyperpigmented disorders are a common complaint in the adult population of all races. First-line topical treatments are usually hydroquinone or topical retinoids, which can cause irritant reactions. The need for better tolerated, yet effective, skin lightening agents that could
Dong-Ho Seo et al.
Applied microbiology and biotechnology, 94(5), 1189-1197 (2012-02-09)
α-Arbutin (α-Ab) is a powerful skin whitening agent that blocks epidermal melanin biosynthesis by inhibiting the enzymatic oxidation of tyrosine and L-3,4-dihydroxyphenylalanine (L-DOPA). α-Ab was effectively synthesized from hydroquinone (HQ) by enzymatic biotransformation using amylosucrase (ASase). The ASase gene from
Hailan Li et al.
Die Pharmazie, 66(4), 306-309 (2011-05-27)
Arbutin is a glycosylated hydroquinone extracted from the bearberry plant (Arctostaphylos species). In the present study, we determined the effects of arbutin on TCCSUP human bladder carcinoma cell proliferation. Arbutin did not exhibit any cytotoxic effects in TCCSUP cells at
Shazia Khan et al.
Natural product communications, 6(11), 1661-1664 (2012-01-10)
A new arbutin derivative, madhuglucoside (1), along with three known arbutin derivatives were isolated from the seeds of Madhuca latifolia in addition to seven other known constituents. Their structures were established on the basis of spectral analysis. Compounds 1a, 2a
Eiman M-E Aboagla et al.
Theriogenology, 76(3), 538-546 (2011-05-03)
Arbutin (4-hydroxyphenyl-glucopyranoside) is a glycosylated hydroquinone present in high concentrations in the leaves of several plants capable of surviving prolonged, extreme dehydration. Two experiments were conducted to determine the effects of arbutin on cryopreservation of goat sperm. In Experiment 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service