Skip to Content
Merck
All Photos(1)

Documents

Y0000672

Iohexol for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-[N-(2,3-Dihydroxypropyl)acetamido]-2,4,6-triiodo-N,N′-bis(2,3-dihydroxypropyl)isophthalamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26I3N3O9
CAS Number:
Molecular Weight:
821.14
Beilstein:
2406632
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

iohexol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

IC1=C(C(NCC(O)CO)=O)C(I)=C(C(NCC(CO)O)=O)C(I)=C1N(CC(CO)O)C(C)=O

InChI

1S/C19H26I3N3O9/c1-8(29)25(4-11(32)7-28)17-15(21)12(18(33)23-2-9(30)5-26)14(20)13(16(17)22)19(34)24-3-10(31)6-27/h9-11,26-28,30-32H,2-7H2,1H3,(H,23,33)(H,24,34)

InChI key

NTHXOOBQLCIOLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Iohexol for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marlijne E Ikink et al.
European radiology, 24(10), 2649-2657 (2014-08-15)
To compare the effectiveness of magnetic resonance-guided high-intensity focused ultrasound (MR-HIFU) with that of uterine artery embolisation (UAE) for treatment of uterine fibroids. Between January 2010 and January 2013, 51 women with symptomatic uterine fibroids underwent MR-HIFU. Follow-up and MR imaging
Aneliya Parvanova et al.
The lancet. Diabetes & endocrinology, 1(1), 19-27 (2014-03-14)
Effective reduction of albuminuria and blood pressure in patients with type 2 diabetes who have nephropathy is seldom achieved with available treatments. We tested the effects of treatment of such patients with daglutril, a combined endothelin-converting enzyme and neutral endopeptidase
Alison G Abraham et al.
Pediatric nephrology (Berlin, Germany), 29(7), 1231-1238 (2014-02-08)
In adults with chronic kidney disease (CKD), protein-energy wasting (PEW) is a risk factor for hospitalization and death. However, PEW in children with CKD is not well characterized or defined. Using data from the Chronic Kidney Disease in Children study
Robert P Jones et al.
AJR. American journal of roentgenology, 204(3), 519-526 (2015-02-26)
OBJECTIVE. The purpose of this article is to evaluate the hypothesis that Alvarado scores of 3 or lower identify adult patients who are unlikely to benefit from CT after appendiceal ultrasound fails to show the appendix and is otherwise normal.
Catrina Hansen et al.
AJR. American journal of roentgenology, 202(4), W357-W364 (2014-03-26)
The purpose of this study was to evaluate the diagnostic performance of a dedicated dual-source dual-energy MDCT (DECT) protocol for the detection of urothelial tumors. A DECT protocol including furosemide and split-bolus contrast injection was used in 69 consecutive patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service