Skip to Content
Merck
All Photos(1)

Key Documents

Y0000663

Methotrexate impurity C

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Methopterine, N10-Methylfolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21N7O6
CAS Number:
Molecular Weight:
455.42
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

methotrexate

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N(Cc2nc3c(nc(nc3O)N)nc2)(C)c1ccc(cc1)C(=O)NC(CCC(=O)O)C(=O)O

InChI

1S/C20H21N7O6/c1-27(9-11-8-22-16-15(23-11)18(31)26-20(21)25-16)12-4-2-10(3-5-12)17(30)24-13(19(32)33)6-7-14(28)29/h2-5,8,13H,6-7,9H2,1H3,(H,24,30)(H,28,29)(H,32,33)(H3,21,22,25,26,31)

InChI key

HLIXOCXUWGDBNP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Methotrexate impurity C EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Muta. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S Godfrey et al.
Lancet (London, England), 336(8712), 392-395 (1990-08-18)
41 (33%) of 123 patients with acute psychiatric disorders (DSM III diagnosis of major depression or schizophrenia) had borderline or definite folate deficiency (red-cell folate below 200 micrograms/l) and took part in a double-blind, placebo-controlled trial of methylfolate, 15 mg
J R Piper et al.
Journal of medicinal chemistry, 29(6), 1080-1087 (1986-06-01)
Evidence indicating that modifications at the 5- and 10-positions of classical folic acid antimetabolites lead to compounds with favorable differential membrane transport in tumor vs. normal proliferative tissue prompted an investigation of 5-alkyl-5-deaza analogues. 2-Amino-4-methyl-3,5-pyridinedicarbonitrile, prepared by hydrogenolysis of its
J M Scott et al.
Lancet (London, England), 2(8242), 337-340 (1981-08-15)
It is suggested that in man the methyl folate trap is a normal physiological response to impending methyl group deficiency resulting from a very low supply of methionine. This decreases cellular S-adenosyl-methionine (SAM), which puts at risk important methylation reactions
R J de Wit et al.
Biochimica et biophysica acta, 930(1), 1-9 (1987-08-19)
The response of guanylate cyclase to addition of extracellular stimuli is well documented. Here we report for the first time the response of guanylate cyclase to removal of stimuli. Three methods were employed to terminate rapidly a stimulus of folic
Enhancement of recovery from psychiatric illness by methylfolate.
T Bottiglieri et al.
Lancet (London, England), 336(8730), 1579-1580 (1990-12-22)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service