Skip to Content
Merck
All Photos(1)

Documents

Y0000621

Terazosin hydrochloride dihydrate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-[(tetrahydro-2-furanyl)carbonyl]piperazine hydrochloride dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25N5O4 · HCl · 2H2O
CAS Number:
Molecular Weight:
459.92
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

terazosin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.Cl.COc1cc2nc(nc(N)c2cc1OC)N3CCN(CC3)C(=O)C4CCCO4

InChI

1S/C19H25N5O4.ClH.2H2O/c1-26-15-10-12-13(11-16(15)27-2)21-19(22-17(12)20)24-7-5-23(6-8-24)18(25)14-4-3-9-28-14;;;/h10-11,14H,3-9H2,1-2H3,(H2,20,21,22);1H;2*1H2

InChI key

NZMOFYDMGFQZLS-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Terazosin hydrochloride dihydrate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie C Lazzaro et al.
Learning & memory (Cold Spring Harbor, N.Y.), 17(10), 489-493 (2010-09-28)
Norepinephrine receptors have been studied in emotion, memory, and attention. However, the role of alpha1-adrenergic receptors in fear conditioning, a major model of emotional learning, is poorly understood. We examined the effect of terazosin, an alpha1-adrenergic receptor antagonist, on cued
Man Liu et al.
Chirality, 24(12), 1047-1050 (2012-08-14)
The purpose of this study was to elucidate the pharmacokinetics of terazosin enantiomers in healthy Chinese male subjects. After a single oral dose of 2-mg terazosin, the plasma concentrations of terazosin enantiomers were measured over the course of 48 h
D Mitropoulos et al.
Andrologia, 45(2), 128-134 (2012-06-27)
Previous studies have shown that alpha1-adrenergic receptor antagonists may alter seminal vesicle contractility and impair fertility in male rats. This study was designed to investigate the effects of terazosin on the catalase expression in the seminal vesicles and the lipid
M R Yadav et al.
Pharmaceutical biology, 50(4), 439-442 (2011-12-06)
The problem of hypertension has gained enormous proportions in the past decade. Multifactorial etiology and complex pathophysiology of the disease has rendered the treatment of the disease a hard task. Sympathetic nervous system and the renin-angiotensin-aldosterone system are primary contributors
Laurel E Ecke et al.
Neuroreport, 23(5), 325-330 (2012-02-18)
The rewarding properties of psychomotor stimulants are traditionally thought to be independent of norepinephrine. Recent findings, however, suggest that local noradrenergic signaling through α1 receptors in the medial prefrontal cortex and the ventral tegmental area - brain regions critically important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service