Skip to Content
Merck
All Photos(1)

Key Documents

Y0000559

Clobetasol propionate

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32ClFO5
CAS Number:
Molecular Weight:
466.97
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clobetasol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl

InChI

1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1

InChI key

CBGUOGMQLZIXBE-XGQKBEPLSA-N

Gene Information

human ... NR3C1(2908)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clobetasol propionate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark G Lebwohl et al.
Journal of the American Academy of Dermatology, 69(3), 385-392 (2013-05-07)
Few clinical trials have evaluated the combination of topical corticosteroids plus systemic therapies for psoriasis. We sought to evaluate efficacy and safety of etanercept plus topical clobetasol propionate (CP) foam versus etanercept monotherapy for treatment of moderate to severe plaque
S R Wiegell et al.
The British journal of dermatology, 171(6), 1487-1492 (2014-07-26)
Photodynamic therapy (PDT) is an effective and established treatment for actinic keratoses (AK) and nonmelanoma skin cancer. The main side-effects of PDT are post-treatment erythema and oedema, and pain during illumination. Severe erythema after PDT enhances the down time associated
Sidharth Sonthalia et al.
International journal of dermatology, 51(11), 1371-1378 (2012-10-17)
Oral lichen planus (OLP) is a common disease of the oral mucosa with worldwide distribution and overall prevalence of 0.5-2.2%. Its etiology remains unclear, although the role of autoimmunity is supported by its association with other autoimmune diseases and the
Leon H Kircik et al.
Journal of drugs in dermatology : JDD, 12(3), 328-334 (2013-04-03)
Chronic hand dermatitis may have a significant detrimental effect on daily home-related and work-related activities, and quality of life (QOL). Clobetasol propionate foam, 0.05%, is indicated for the treatment of inflammatory and pruritic manifestations of corticosteroid-responsive dermatoses in patients aged
Steven R Feldman et al.
American journal of clinical dermatology, 10(6), 397-406 (2009-10-15)
Ultrapotent topical corticosteroids are the mainstay of psoriasis treatment, used either alone or in combination with a topical vitamin D analog. Traditionally used in an ointment vehicle for psoriasis, clobetasol propionate 0.05% is also available in spray, foam, lotion, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service