Skip to Content
Merck
All Photos(1)

Key Documents

Y0000378

Mitomycin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Mitomycin C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N4O5
CAS Number:
Molecular Weight:
334.33
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

mitomycin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N21[C@]([C@H]4N[C@H]4C2)([C@@H](C3=C1C(=O)C(=C(C3=O)N)C)COC(=O)N)OC

InChI

1S/C15H18N4O5/c1-5-9(16)12(21)8-6(4-24-14(17)22)15(23-2)13-7(18-13)3-19(15)10(8)11(5)20/h6-7,13,18H,3-4,16H2,1-2H3,(H2,17,22)/t6-,7+,13+,15-/m1/s1

InChI key

NWIBSHFKIJFRCO-WUDYKRTCSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Mitomycin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral - Carc. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ingeborg B Fraunholz et al.
Diseases of the colon and rectum, 57(4), 423-431 (2014-03-13)
Despite the increasing evidence for chemoradiotherapy as standard treatment for anal cancer in patients with HIV infection, there is still some uncertainty regarding increased toxicity and adverse effects on the immune status. We report the clinical outcome of 5-fluorouracil/mitomycin C-based
Rebecca Kleinerman et al.
Journal of drugs in dermatology : JDD, 12(6), 701-703 (2013-07-11)
Mitomycin C (MMC) is an antineoplastic antibiotic that has been used off-label in the treatment of hypertrophic scars and keloids. Herein we report our successful use of this agent in a patient with sternal keloids refractory to other means of
[The mechanism and progress of topical application of mitomycin C as an adjunctive therapy for benign airway stenosis].
Nan Chen et al.
Zhonghua jie he he hu xi za zhi = Zhonghua jiehe he huxi zazhi = Chinese journal of tuberculosis and respiratory diseases, 36(8), 592-594 (2013-11-21)
Jianfeng Xiang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 3517-3524 (2013-12-07)
Hepatocellular carcinoma (HCC) is one of the most aggressive malignancies in the world whose chemoprevention became increasingly important in HCC treatment. Although the anticancer effects of asparagus constituents have been investigated in several cancers, its effects on hepatocellular carcinoma have
D Hompes et al.
Journal of surgical oncology, 109(6), 527-532 (2014-01-01)
Oxaliplatin and Mitomycin C (MMC) are both suitable as intraperitoneal chemotherapy agents in HIPEC for peritoneal carcinomatosis (PC) of colorectal cancer (CRC). Patient cohorts from two different HIPEC-centers underwent cytoreductive surgery and HIPEC with Oxaliplatin (39 patients) and MMC (56

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service