Skip to Content
Merck
All Photos(1)

Key Documents

Y0000364

Clioquinol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Chloro-7-iodo-8-quinolinol, 5-Chloro-8-hydroxy-7-iodoquinoline, Clioquinol, Iodochlorhydroxyquin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5ClINO
CAS Number:
Molecular Weight:
305.50
Beilstein:
153637
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

clioquinol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1c(I)cc(Cl)c2cccnc12

InChI

1S/C9H5ClINO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H

InChI key

QCDFBFJGMNKBDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Clioquinol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A D Schimmer
Current cancer drug targets, 11(3), 325-331 (2011-01-21)
Clioquinol (5-chloro-7-iodo-quinolin-8-ol) was used in the 1950's-1970's as an oral anti-parasitic agent. More recently, studies have demonstrated that Clioquinol displays preclinical efficacy in the treatment of malignancy. Its anti-cancer activity relates, at least in part, to its ability to inhibit
Masato Katsuyama et al.
Toxicology, 299(1), 55-59 (2012-05-26)
Clioquinol, a Cu²⁺/Zn²⁺/Fe²⁺ chelator/ionophor, was used extensively in the mid 1900s as an amebicide for treating indigestion and diarrhea. It was eventually withdrawn from the market because of a link to subacute myelo-optic neuropathy (SMON) in Japan. The pathogenesis of
Aaron D Schimmer et al.
Clinical lymphoma, myeloma & leukemia, 12(5), 330-336 (2012-06-12)
Clioquinol is a small-molecule metal ionophore that inhibits the proteasome through a metal-dependent mechanism. Here, we report a phase I study of clioquinol in patients with refractory hematologic malignancies. Neuropathy and abdominal pain were dose-limiting toxicities. Minimal pharmacodynamic effects were
Kouichi Yoshinari et al.
Toxicology letters, 206(2), 229-233 (2011-08-09)
Clioquinol, originally marketed as an oral intestinal amebicide, was widely used for multiple intestinal disorders. Its use as an oral agent was, however, discontinued because of its possible association with subacute myelo-optico-neuropathy or SMON. Meanwhile, its use for neurodegenerative diseases
Tao Wang et al.
Journal of Alzheimer's disease : JAD, 29(3), 549-559 (2012-01-25)
Metal dyshomeostasis in the brain helps promote amyloid-β (Aβ) deposition in Alzheimer's disease (AD). Therefore, targeting the interactions between metal and Aβ is a potential therapeutic approach for AD. The metal chelator, clioquinol (CQ), is thought to reduce Aβ deposits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service