Skip to Content
Merck
All Photos(3)

Key Documents

W292912

Sigma-Aldrich

Isopropyl alcohol

greener alternative

natural, ≥98%, FG

Synonym(s):

2-Propanol, sec-Propyl alcohol, IPA, Isopropanol, Isopropyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOH
CAS Number:
Molecular Weight:
60.10
FEMA Number:
2929
Beilstein:
635639
EC Number:
Council of Europe no.:
4146
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
2.079
NACRES:
NA.21
grade:
FG
Fragrance grade
Halal
Kosher
natural
biological source:
botanical
food allergen:
no known allergens

biological source

botanical

Quality Level

300
400

grade

FG
Fragrance grade
Halal
Kosher
natural

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 172.515

vapor density

2.1 (vs air)

vapor pressure

33 mmHg ( 20 °C)
44 mmHg ( 25 °C)

Assay

≥98%

autoignition temp.

750 °F

expl. lim.

2.0-12.7 %, 93 °C

greener alternative product characteristics

Less Hazardous Chemical Syntheses
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

refractive index

n20/D 1.377 (lit.)

bp

82 °C (lit.)

mp

−89.5 °C (lit.)

density

0.785 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

greener alternative category

Organoleptic

alcohol; ethereal

SMILES string

CC(C)O

InChI

1S/C3H8O/c1-3(2)4/h3-4H,1-2H3

InChI key

KFZMGEQAYNKOFK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product is a Biobased products, showing key improvements in Green Chemistry Principles “Less Hazardous Chemical Syntheses” and “Use of Renewable Feedstock”.

Application


  • Different solvents and organic modifiers for the control of crystallographic parameters in nano-crystallite hydroxyapatite for amplification of photocatalytic activity.: This study demonstrates how isopropyl alcohol, used as a solvent, can control the structural properties of hydroxyapatite nanocrystals, enhancing their photocatalytic performance in environmental applications (Kawsar et al., 2024).

  • Preparation of a sustainable magnetic sorbent for the extraction and preconcentration of progestogens in natural water samples.: The study utilizes isopropyl alcohol in the synthesis of a novel, environmentally friendly sorbent for water purification processes, addressing the crucial need for removing contaminants from water (Aguinaga Martínez et al., 2024).

  • Rational synthesis of methylsilsesquioxane aerogels addressing thermal load and compression recovery issues in Li-ion batteries.: This research highlights the use of isopropyl alcohol in synthesizing advanced materials that improve the safety and efficiency of lithium-ion batteries, a critical component for sustainable energy solutions (Li et al., 2024).

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

53.6 °F - closed cup

Flash Point(C)

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatrice Muzzi et al.
Nanoscale, 12(26), 14076-14086 (2020-06-26)
The development of reproducible protocols to synthesize hard/soft nano-heterostructures (NHSs) with tailored magnetic properties is a crucial step to define their potential application in a variety of technological areas. Thermal decomposition has proved to be an effective tool to prepare
Ivan V Semenyuta et al.
International journal of molecular sciences, 22(2) (2021-01-13)
Online Chemical Modeling Environment (OCHEM) was used for QSAR analysis of a set of ionic liquids (ILs) tested against multi-drug resistant (MDR) clinical isolate Acinetobacter baumannii and Staphylococcus aureus strains. The predictive accuracy of regression models has coefficient of determination
Shuli Yin et al.
Nanoscale, 12(27), 14863-14869 (2020-07-08)
The development of doped noble metal catalysts with nonmetal elements to improve the catalytic performance toward the oxygen reduction reaction (ORR) is significant for proton exchange membrane fuel cell technology. Here, we report a one-pot for dual-nonmetal-doping strategy for the
Heba H El-Maghrabi et al.
Journal of colloid and interface science, 587, 457-466 (2021-01-01)
Cost-effective, active and stable electrocatalysts are crucial for hydrogen production via electrocatalytic water splitting. Here, we describe the preparation of novel nanofibers (NF) made of Ni/Gd2O3/NiO heterostructures by electrospinning. The fabricated materials showed high electrocatalytic performance for hydrogen evolution reaction
Justyna Odrobińska et al.
Pharmaceutics, 12(12) (2020-12-09)
The in vitro biochemical evaluation of the applicability of polymers carrying active substances (micelles and conjugates) was carried out. Previously designed amphiphilic graft copolymers with retinol or 4-n-butylresorcinol functionalized polymethacrylate backbone and poly(ethylene glycol) (PEG) side chains that included Janus-type

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service