Skip to Content
Merck
All Photos(1)

Documents

T1350000

Thymol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Isopropyl-5-methylphenol, 5-Methyl-2-(1-methylethyl)phenol, 5-Methyl-2-isopropylphenol, IPMP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
2-[(CH3)2CH]C6H3-5-(CH3)OH
CAS Number:
Molecular Weight:
150.22
Beilstein:
1907135
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

Agency

EP Reference Standard

vapor pressure

1 mmHg ( 64 °C)

API family

thymol

manufacturer/tradename

EDQM

bp

232 °C (lit.)

mp

48-51 °C (lit.)

density

0.965 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)c1ccc(C)cc1O

InChI

1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3

InChI key

MGSRCZKZVOBKFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Thymol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

240.8 °F - closed cup

Flash Point(C)

116 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Kohlert et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 767(1), 11-18 (2002-02-28)
A reliable and sensitive method was developed for determination of thymol in human plasma by automated headspace solid-phase microextraction (SPME). After enzymatic cleavage of thymol sulfate thymol was extracted by a 65 microm polydimethylsiloxane-divinylbenzene crimped fiber (Supelco) after addition of
Hasan Mahmud et al.
Cardiovascular research, 98(1), 37-46 (2013-01-24)
Suicidal death of erythrocytes (eryptosis) is characterized by cell shrinkage and exposure of phosphatidylserine (PS) residues at the cell surface. Excessive eryptosis may lead to anaemia. We aimed to study the role of eryptosis in heart failure (HF)-associated anaemia. We
R Attin et al.
Archives of oral biology, 48(7), 503-509 (2003-06-12)
The objective of the present prospective trial was to compare the efficacy of differently concentrated chlorhexidine varnishes (EC40) = 40% chlorhexidine and Cervitec = 1% chlorhexidine + 0.1% thymol) on levels of Mutans streptococci (ms), lactobacilli (lb) and plaque formation
María C Elissondo et al.
Acta tropica, 125(3), 251-257 (2012-12-25)
Scolicidal solutions remain indispensable in the treatment of hydatid cyst disease. Properties of an ideal solution would be inexpensiveness and the promotion of a rapid and complete scolicidal effect with an absence of local and systemic side effects. From this
Hiroko Ohwada et al.
Research in developmental disabilities, 34(1), 650-655 (2012-11-06)
As the life expectancy of people with intellectual disability (ID) increases, it is becoming necessary to understand factors affecting survival. However, predictors that are typically assessed among healthy people have not been examined. Predictors of all-cause mortality, including blood, urine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service