Skip to Content
Merck
All Photos(1)

Documents

R0630000

Riboflavin sodium phosphate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Riboflavin 5′-monophosphate sodium salt, Riboflavin Sodium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N4NaO9P
CAS Number:
Molecular Weight:
478.33
Beilstein:
4106529
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP

API family

riboflavin

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Na+].Cc1cc2N=C3C(=O)NC(=O)N=C3N(C[C@H](O)[C@H](O)[C@H](O)COP(O)([O-])=O)c2cc1C

InChI

1S/C17H21N4O9P.Na/c1-7-3-9-10(4-8(7)2)21(15-13(18-9)16(25)20-17(26)19-15)5-11(22)14(24)12(23)6-30-31(27,28)29;/h3-4,11-12,14,22-24H,5-6H2,1-2H3,(H,20,25,26)(H2,27,28,29);/q;+1/p-1/t11-,12+,14-;/m0./s1

InChI key

OHSHFZJLPYLRIP-BMZHGHOISA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Riboflavin sodium phosphate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brian C Smith et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(38), E3577-E3586 (2013-09-05)
Nitric oxide (NO) produced by NO synthase (NOS) participates in diverse physiological processes such as vasodilation, neurotransmission, and the innate immune response. Mammalian NOS isoforms are homodimers composed of two domains connected by an intervening calmodulin-binding region. The N-terminal oxidase
Galina Melman et al.
Biochimica et biophysica acta, 1830(10), 4669-4674 (2013-06-04)
Extensive in-vitro studies have focused on elucidating the mechanism of iron uptake and mineral core formation in ferritin. However, despite a plethora of studies attempting to characterize iron release under different experimental conditions, the in-vivo mobilization of iron from ferritin
Sarah Raffelberg et al.
Biological chemistry, 394(11), 1517-1528 (2013-07-06)
Flavin-binding light, oxygen, and voltage (LOV) domains are UVA/blue-light-sensing protein units that form a reversible flavin mononucleotide-cysteine adduct upon light induction. In their dark-adapted state, LOV domains exhibit the typical spectral features of fully oxidized riboflavin derivatives. A survey on
Jinrok Oh et al.
Organic letters, 15(6), 1210-1213 (2013-03-07)
A new application of a fluorescent sensor (PyDPA) for the discrimination of redox-responsible molecules is reported. Nicotinamide adenine dinucleotide/nicotinamide adenine dinucleotide phosphate (NAD(+)/NADP(+)) and flavin mononucleotide/flavin adenine dinucleotide (FMN/FAD) were differentiated by means of ratiometric fluorescence change from excimer-monomer equilibrium
Jong-Chin Lin et al.
Biophysical journal, 103(11), 2320-2330 (2013-01-04)
Riboswitches, structured elements in the untranslated regions of messenger RNAs, regulate gene expression by binding specific metabolites. We introduce a kinetic network model that describes the functions of riboswitches at the systems level. Using experimental data for flavin mononucleotide riboswitch

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service